Skip to Content
Merck
All Photos(2)

Key Documents

RAB0242

Sigma-Aldrich

Human IgG1 ELISA Kit

for serum, plasma, conditioned medium, urine

Synonym(s):

IgG1, Immunoglobulin G1

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$918.00

$918.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$918.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$918.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type conditioned medium
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 50 pg/mL
standard curve range: 0.05-40 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IGHG1(3500)

General description

The Human IgG1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IgG1 in serum and plasma.

Immunogen

Recombinant Human IGG1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Immunoglobulin heavy constant gamma 1 (IGHG1), or IgG1, expressed in pancreatic cancer cells might be implicated in tumor cell proliferation and immune evasion mechanisms.[1] siRNA targeted silencing of IGHG1 retards cell growth and induces apoptosis in human prostate cancer cell.[2]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Roxanne Collin et al.
Journal of immunology (Baltimore, Md. : 1950), 193(7), 3503-3512 (2014-08-29)
Autoimmune diseases result from a break in immune tolerance. Various mechanisms of peripheral tolerance can protect against autoimmunity, including immunoregulatory CD4(-)CD8(-) double-negative (DN) T cells. Indeed, we have previously shown that diabetes-prone mouse strains exhibit a low proportion of DN
Bin Pan et al.
Molecular biology reports, 40(1), 27-33 (2012-11-03)
To investigate the immunoglobulin G (IgG) expression in prostate cancer cell lines and explore the effects of IGHG1 gene knockdown on PC3 cell growth and apoptosis. Flow cytometry, qPCR and western blot were used to demonstrate IgG expression in prostate
Xiaoyan Li et al.
Pancreas, 40(5), 753-761 (2011-06-10)
To investigate the expression of Igγ-1 chain C region (IGHG1) in human pancreatic carcinomas and determine the biological function of IGHG1 expression in immune evasion mechanisms. Comparative proteomic analysis was used to detect the differential expression of IGHG1 in human
Joseph R Brady et al.
Biotechnology and bioengineering, 117(2), 543-555 (2019-10-28)
Komagataella phaffii, also known as Pichia pastoris, is a common host for the production of biologics and enzymes, due to fast growth, high productivity, and advancements in host engineering. Several K. phaffii variants are commonly used as interchangeable base strains
Arjan van der Flier et al.
PloS one, 10(4), e0124930-e0124930 (2015-04-24)
We recently developed a longer lasting recombinant factor VIII-Fc fusion protein, rFVIIIFc, to extend the half-life of replacement FVIII for the treatment of people with hemophilia A. In order to elucidate the biological mechanism for the elongated half-life of rFVIIIFc

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service