Skip to Content
Merck
All Photos(2)

Key Documents

RAB0079

Sigma-Aldrich

Human MCP-2 / CCL8 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$918.00

$918.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 KIT
$918.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$918.00


Estimated to ship on28 May 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 0.82-200 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL8(6355)

General description

The Human MCP-2 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MCP-2 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human MCP2

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca A Flaherty et al.
PloS one, 14(9), e0222910-e0222910 (2019-09-20)
Group B Streptococcus (GBS) is an opportunistic pathogen that causes preterm birth and neonatal disease. Although GBS is known to exhibit vast diversity in virulence across strains, the mechanisms of GBS-associated pathogenesis are incompletely understood. We hypothesized that GBS strains
Jie Ji et al.
Photodiagnosis and photodynamic therapy, 26, 235-243 (2019-03-25)
Antitumor immunity induced by photodynamic therapy (PDT) is believed to depend on the degree of local and systemic inflammation. The recruitment of leukocytes, in particular by the chemokine CCL8, to the sites of tissue damage has been strongly associated with
Anna Chiarini et al.
Cells, 9(6) (2020-06-06)
Available evidence shows that human cortical neurons' and astrocytes' calcium-sensing receptors (CaSRs) bind Amyloid-beta (Aβ) oligomers triggering the overproduction/oversecretion of several Alzheimer's disease (AD) neurotoxinseffects calcilytics suppress. We asked whether AβCaSR signaling might also play a direct pro-neuroinflammatory role in
Tiyash Parira et al.
Scientific reports, 7(1), 11236-11236 (2017-09-13)
Previous reports on epigenetic mechanisms involved in alcohol abuse have focus on hepatic and neuronal regions, leaving the immune system and specifically monocyte-derived dendritic cells (MDDCs) understudied. Our lab has previously shown histone deacetylases are modulated in cells derived from
Yanling Gou et al.
Human reproduction (Oxford, England), 34(4), 646-658 (2019-03-07)
How is the activation of estrogen receptor β (ERβ) in endometriotic stromal cells (ESCs) involved in macrophage recruitment to promote the pathogenesis of endometriosis? ERβ modulates the production of C-C motif chemokine ligand 2 (CCL2) via nuclear factor kappa-light-chain-enhancer of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service