Key Documents
RA2VC-EP
Rotor for Eppendorf® Vacufuge® Plus Concentrator
swing-out rotor, A-2-VCm non aerosol tight
About This Item
UNSPSC Code:
41103909
Product RA2VC-EP is not currently sold in your country. Contact Technical Service
feature
skirt
to hold 2 x MTP or skirted PCR plates (max height 27 mm)
to hold 2 x deepwell plates (max height 27 mm)
A-2-VCm non aerosol tight
swing-out rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822241 (North America)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)