Skip to Content
Merck
All Photos(1)

Key Documents

R9633

Sigma-Aldrich

D-(−)-Ribose

BioReagent, suitable for plant cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723081
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.56

product line

BioReagent

technique(s)

cell culture | plant: suitable

mp

88-92 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@@H](O)C([H])=O

InChI

1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5-/m0/s1

InChI key

PYMYPHUHKUWMLA-LMVFSUKVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy G Briggs et al.
Trends in plant science, 16(7), 372-380 (2011-04-13)
Poly(ADP-ribose) polymerases (PARPs) and poly(ADP-ribose) glycohydrolases (PARGs) are the main enzymes responsible for the post-translational modification known as poly(ADP-ribosyl)ation. These enzymes play important roles in genotoxic stress tolerance and DNA repair, programmed cell death, transcription, and cell cycle control in
Raman K Sharma et al.
Bioorganic & medicinal chemistry, 20(23), 6821-6830 (2012-10-27)
A series of peracetylated O-aryl α,β-d-ribofuranosides have been synthesized and an efficient biocatalytic methodology has been developed for the separation of their anomers which was otherwise almost impossible by column chromatographic or other techniques. The incubation of 2,3,5-tri-O-acetyl-1-O-aryl-α,β-d-ribofuranoside with Lipozyme®
Anders Virtanen et al.
Critical reviews in biochemistry and molecular biology, 48(2), 192-209 (2013-03-19)
Deadenylation of eukaryotic mRNA is a mechanism critical for mRNA function by influencing mRNA turnover and efficiency of protein synthesis. Here, we review poly(A)-specific ribonuclease (PARN), which is one of the biochemically best characterized deadenylases. PARN is unique among the
Xiang-Guo Li et al.
Chemical communications (Cambridge, England), 49(35), 3682-3684 (2013-03-29)
Peptide glycosylation with 5-deoxy-5-[(18)F]fluororibose was translated into preclinical settings. The novel (18)F-labeled Siglec-9 peptide was produced using an automated synthesis procedure. The (18)F-labeled Siglec-9 peptide showed favorable binding in the animal model of inflammation in vivo.
Carine Baraguey et al.
Organic & biomolecular chemistry, 11(16), 2638-2647 (2013-03-05)
The pivaloyloxymethyl (PivOM) group is a biolabile 2'-O-ribose protection that is under development in a prodrug-based approach for siRNA applications. Besides an expected cellular uptake, nucleic acid sequences carrying PivOM showed also increased nuclease resistance and, in most cases, an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service