Skip to Content
Merck
All Photos(2)

Key Documents

R8750

Sigma-Aldrich

Methyl ricinoleate

≥99% (GC)

Synonym(s):

(R)-12-Hydroxy-cis-9-octadecenoic acid methyl ester, Methyl 12-hydroxyoleate, Ricinoleic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$76.40
1 G
$219.00

$76.40


Available to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$76.40
1 G
$219.00

About This Item

Empirical Formula (Hill Notation):
C19H36O3
CAS Number:
Molecular Weight:
312.49
Beilstein:
6132055
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

$76.40


Available to ship on10 April 2025Details


Request a Bulk Order

Assay

≥99% (GC)

form

liquid

functional group

ester

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(OC)CCCCCCC/C=C\C[C@H](O)CCCCCC

InChI

1S/C19H36O3/c1-3-4-5-12-15-18(20)16-13-10-8-6-7-9-11-14-17-19(21)22-2/h10,13,18,20H,3-9,11-12,14-17H2,1-2H3/b13-10-/t18-/m1/s1

InChI key

XKGDWZQXVZSXAO-ADYSOMBNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Endrizzi et al.
Journal of basic microbiology, 35(5), 285-292 (1995-01-01)
The capacity of several strains of yeasts to do the bioconversion of methyl ricinoleate into gamma-decalactone, was studied in a medium containing this methylic ester of fatty acid as sole carbon source. Amongst the strains which are able to do
David S Wunschel et al.
The Analyst, 137(9), 2077-2085 (2012-03-15)
The investigation of crimes involving chemical or biological agents is infrequent, but presents unique analytical challenges. The protein toxin ricin is encountered more frequently than other agents and is found in the seeds of Ricinus communis, commonly known as the
Y Waché et al.
Letters in applied microbiology, 30(3), 183-187 (2000-04-04)
Size of methyl ricinoleate droplets during biotransformation into gamma-decalactone by Yarrowia lipolytica was measured in both homogenized and non-homogenized media. In non-homogenized but shaken medium, droplets had an average volume surface diameter d32 of 2.5 microm whereas it was 0.7
J T Lin et al.
Lipids, 35(5), 481-486 (2000-07-25)
We have examined the role of 2-oleoyl-PE (phosphatidylethanolamine) in the biosynthesis of triacylglycerols (TAG) by castor microsomes. In castor microsomal incubation, the label from 14C-oleate of 1-palmitoyl-2-[1-(14)C]oleoyl-sn-glycero-3-phosphoethanolamine is incorporated into TAG containing ricinoleate. The enzyme characteristics, such as optimal pH
M S Jie et al.
Lipids, 30(2), 115-120 (1995-02-01)
Using a combination of selective irradiation 1H nuclear magnetic resonance experiments and two-dimensional 1H-13C correlation spectroscopy spectral analysis of homoallylic and bis homoallylic substituted (azido, acetoxy, chloro and oxo) fatty ester derivatives, the carbon shifts of the ethylenic carbon atoms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service