Skip to Content
Merck
All Photos(3)

Key Documents

R5780

Sigma-Aldrich

Anti-Rab4 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HRES-1/RAB4, Anti-RAB4A, Anti-RAS-associated protein RAB4

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$737.00

$737.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$737.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$737.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~24 kDa

species reactivity

human, rat, mouse

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using mouse 3T3 and rat NRK cells fixed and permeabilized with 4% paraformaldehyde followed by 0.1% and 0.4% saponin, respectively.
western blot: 2-5 μg/mL using whole extract of human HEK-293T cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB4A(5867)
mouse ... Rab4a(19341)
rat ... Rab4a(25532)

General description

Rab4 is a GTPase , that is located in early and recycling endosomes. It belongs to Rab family and is located on human chromosome 1q42. In rat testis, Rab4 is found in sertoli and germ cells.
There are two highly homologous RAB4 isoforms, RAB4A and RAB4B, encoded by two separate genes (RAB4A and RAB4B). RAB4A and RAB4B are localized in the same cellular compartments and are believed to have similar or identical functions in recycling.

Application

The antibody can be used in several immunochemical techniques including immunoblotting (24 kDa) and immunofluorescence.

Biochem/physiol Actions

Rab4 regulates endocytic recycling in a cell cycle dependent fashion. Rab4 is associated with early endosomes and recycling endosomes and regulates the recycling of membranes and proteins from these compartments back to the plasma membrane. Rab4 interacts with several Rab4 effector proteins like Rabip4, that regulates integrin trafficking and is involved in the migration of NIH 3T3 fibroblasts.
Rab4a controls integrin trafficking throughout cell adhesion. It helps in the quick reprocess of receptors from early endosomes to the cell surface. It also controls the secretion of procathepsin L in human melanoma cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rab4A GTPase catenin interactions are involved in cell junction dynamics in the testis
Mruk DD, et al.
Journal of Andrology, 28(5), 742-754 (2007)
TBC1D16 is a Rab4A GTPase activating protein that regulates receptor recycling and EGF receptor signaling
Goueli BS, et al.
Proceedings of the National Academy of Sciences of the USA, 109(39), 15787-15792 (2012)
Expression of RAB4B, a protein governing endocytic recycling, is co-regulated with MHC class II genes
Krawczyk M, et al.
Nucleic Acids Research, 35(2), 595-605 (2006)
The Rab4A effector protein Rabip4 is involved in migration of NIH 3T3 fibroblasts
Vukmirica J, et al.
The Journal of Biological Chemistry, 281(47), 36360-36368 (2006)
Procathepsin L secretion, which triggers tumour progression, is regulated by Rab4a in human melanoma cells
Barbarin A and Frade R
The Biochemical Journal, 437(1), 97-107 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service