Skip to Content
Merck
All Photos(1)

Key Documents

R311-500

Sigma-Aldrich

Rat Smooth Muscle Cell Growth Medium (500 ml)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$269.00

$269.00


Estimated to ship on17 May 2025



Select a Size

Change View
1 EACH
$269.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$269.00


Estimated to ship on17 May 2025


form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media

Application

Rat Smooth Muscle Cell Growth Medium (500 ml) has been used to culture rat aortic smooth muscle cells.[1]

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Zhang et al.
Materials science & engineering. C, Materials for biological applications, 115, 111090-111090 (2020-07-01)
The in-stent restenosis (ISR) often happens after the implantation of metal stents, including both bare metal stents (BMSs) and drug-eluting stents (DESs). Drug release from DESs could reduce significantly the occurrence of ISR but also suppress the revascularization and cause

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service