Skip to Content
Merck
All Photos(1)

Key Documents

R1783

Sigma-Aldrich

Rhodblock 1a

≥98% (HPLC)

Synonym(s):

(4,5-Dihydro-3,5-diphenyl-1H-pyrazol-1-yl)-2-furanyl-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$542.00

$542.00


Estimated to ship on17 May 2025



Select a Size

Change View
25 MG
$542.00

About This Item

Empirical Formula (Hill Notation):
C20H16N2O2
CAS Number:
Molecular Weight:
316.35
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$542.00


Estimated to ship on17 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

O=C(N1N=C(CC1c2ccccc2)c3ccccc3)c4ccco4

InChI

1S/C20H16N2O2/c23-20(19-12-7-13-24-19)22-18(16-10-5-2-6-11-16)14-17(21-22)15-8-3-1-4-9-15/h1-13,18H,14H2

InChI key

VEIXLXIVAMCBLZ-UHFFFAOYSA-N

Biochem/physiol Actions

Rhodblock 1a is an inhibitor of the Rho Kinase pathway.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service