Skip to Content
Merck
All Photos(1)

Key Documents

R1656

Sigma-Aldrich

RAF1 (EE) (306-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CMD1NN, NS5, RAF proto-oncogene serine/threonine-protein kinase, cRaf

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$664.00

$664.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$664.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$664.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

fermentation/recombinant

Quality Level

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

5100-6900 nmol/min·mg

mol wt

~63 kDa

manufacturer/tradename

Precision

technique(s)

cell based assay: suitable

solubility

water: soluble

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RAF1(5894)

General description

Research area: Cell Signaling

Rapidly Accelerated Fibrosarcoma (RAF) belongs to receptor tyrosine kinase (RTK) family. RAF exists as three isoforms A, B and C-RAF or RAF-1. [1] RAF1 links Ras family members with mitogen-activated protein kinase/extracellular signal-regulated kinase (MAPK/ERK) signaling cascade.[2] The RAF-1 gene is mapped to the human chromosome at 3p25.2.[3]

Biochem/physiol Actions

Rapidly Accelerated Fibrosarcoma 1(RAF1), a member of MAP kinase kinase kinase (MAP3K) family plays a vital role in cell signaling by directly interacting with GTPases and functions downstream in their signaling pathway.[4] RaF1 is activated by Ras in the presence of growth-promoting signals, leading to the activation of mitogen-activated protein kinase kinase (MEK1/MEK2) and extracellular signal-regulated protein kinases 1 and 2 (ERK1/ERK2) downstream. [5] The activated ERK proteins control gene expression related to cell division, apoptosis, cell differentiation, and cell migration.[4] The gene is mutated or upregulated in prostate and bladder cancer.[5] Also, mutations are associated with Noonan syndrome, which leads to various physical abnormalities like craniofacial dysmorphisms, ocular, and musculoskeletal irregularities, neurocognitive impairment, and cardiac malformations.[4]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAF1 deficiency causes a lethal syndrome that underscores RTK signaling during embryogenesis
Wong S, et al.
EMBO Molecular Medicine, 15(5), e17078-e17078 (2023)
Benjamin R Nixon et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(1), 1138-1150 (2018-08-15)
Raf1/c-Raf is a well-characterized serine/threonine-protein kinase that links Ras family members with the MAPK/ERK signaling cascade. We have identified a novel splice isoform of human Raf1 that causes protein truncation and loss of the C-terminal kinase domain (Raf1-tr). We found
Olafur Andri Stefansson et al.
Breast cancer research : BCR, 13(3), 309-309 (2011-06-16)
Members of the Polycomb-group (PcG) family of proteins, including EZH2 (enhancer of zeste homolog 2), are involved in establishing epigenetic silencing of developmental genes in adult and embryonic stem cells, and their deregulation has been implicated in cancer. In a
Raf kinases: function, regulation and role in human cancer
Leicht DT, et al.
Biochimica et Biophysica Acta, 1773(8), 1196-1212 (2007)
Vertical pathway targeting in cancer therapy
Shahbazian D, et al.
Advances in Pharmacology, 65, 1-26 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service