Skip to Content
Merck
All Photos(1)

Key Documents

R116-500

Sigma-Aldrich

Rat Fibroblast Growth Medium (500 ml)

Synonym(s):

Fibroblast Growth Medium

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$202.00

$202.00


Estimated to ship on08 April 2025



Select a Size

Change View
1 EACH
$202.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$202.00


Estimated to ship on08 April 2025


form

liquid

Quality Level

shelf life

6 mo.

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media

Application

Rat Fibroblast Growth Medium (500 ml) has been used to culture rat dermal fibroblast cells[1] and primary rat lung fibroblast cells.[2]

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruochong Fei et al.
Macromolecular materials and engineering, 301(8), 935-943 (2017-05-23)
A self-cleaning membrane that periodically rids itself of attached cells to maintain glucose diffusion could extend the lifetime of implanted glucose biosensors. Herein, we evaluate the functionality of thermoresponsive double network (DN) hydrogel membranes based on poly(N-isopropylacrylamide) (PNIPAAm) and an
Sana Bahri et al.
PloS one, 12(9), e0184368-e0184368 (2017-09-07)
Pulmonary fibrosis is characterized by over-population and excessive activation of fibroblasts and myofibroblasts disrupting normal lung structure and functioning. Rosemary extract rich in carnosic acid (CA) and rosmarinic acid (RA) was reported to cure bleomycin-(BLM)-induced pulmonary fibrosis. We demonstrate that

Protocols

Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service