Skip to Content
Merck
All Photos(1)

Key Documents

R107

Sigma-Aldrich

Ro 41-1049 hydrochloride

solid

Synonym(s):

N-(2-Aminoethyl)-5-(3-fluorophenyl)-4-thiazolecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12FN3OS · HCl
CAS Number:
Molecular Weight:
301.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

white

solubility

H2O: soluble

SMILES string

Cl[H].NCCNC(=O)c1ncsc1-c2cccc(F)c2

Gene Information

human ... MAOA(4128)

Biochem/physiol Actions

Selective, reversible, orally-active MAO-A inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Heeringa et al.
Journal of neural transmission (Vienna, Austria : 1996), 104(6-7), 593-603 (1997-01-01)
The effect of enzyme-inhibiting adjuvants on L-DOPA + benserazide-induced contralateral turning in unilateral 6-hydroxydopamine (6-OHDA)-lesioned rats was studied. Both the number of turns and the duration of turning were examined. Inhibition of MAO-A with 10 mg/kg Ro 41-1049 increased both
A Ozaita et al.
British journal of pharmacology, 121(5), 901-912 (1997-07-01)
1. I2-Imidazoline sites ([3H]-idazoxan binding) have been identified on monoamine oxidase (MAO) and proposed to modulate the activity of the enzyme through an allosteric inhibitory mechanism (Tesson et al., 1995). The main aim of this study was to assess the
M H Fernandes et al.
The Journal of pharmacology and experimental therapeutics, 255(3), 1309-1313 (1990-12-01)
The present study has examined the effects of two selective inhibitors of monoamine oxidase (MAO) type A and B, respectively Ro 41-1049 and Ro 19-6327, on the deamination of newly synthesized dopamine (DA) in kidney slices incubated with exogenous L-3,4-dihydroxyphenylalanine
J Saura et al.
Life sciences, 59(16), 1341-1349 (1996-01-01)
Localization of monoamine oxidases (MAO) A and B and beta-adrenoceptors, was studied in aged human peripheral tissues (age 68-80 years) by quantitative autoradiography. The tissues analyzed were heart, lung, liver, kidney, spleen and duodenum. [3H]Ro41-1049 and [3H]lazabemide, two recently characterized
J Saura et al.
Neurobiology of aging, 18(5), 497-507 (1997-12-09)
Variations of monoamine oxidases (MAO) A and B were studied during aging in 27 human subjects (age range 17-93 years) in 18 brain structures of temporal cortex, frontal gyrus, hippocampal formation, striatum, cerebellum, and brainstem. [3H]Ro41-1049 and [3H]lazabemide were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service