Skip to Content
Merck
All Photos(1)

Documents

R0281

Sigma-Aldrich

RNF11 human

recombinant, expressed in E. coli (histidine-tagged), ≥90% (SDS-PAGE), buffered aqueous solution

Synonym(s):

CGI-123, MGC51169, RING Finger Protein 11, SID1669

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

recombinant

expressed in E. coli (histidine-tagged)

Assay

≥90% (SDS-PAGE)

form

buffered aqueous solution

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... RNF11(26994)

Biochem/physiol Actions

RNF11 (RING Finger Protein 11) is a RING E3 ubiquitin ligase with potential to mediate ubiquitination and subsequent proteolysis. RNF11 binds numerous proteins in a variety of cellular pathways, including TGFβ and EGFR signaling. RNF11 expression is elevated in breast and prostate cancer. RNF11 may represent an important target for novel cancer therapies.

Physical form

Solution in 50 mM Tris, pH 7.5, 150 mM NaCl, 1 mM DTT, and 30% glycerol (w/v).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael K Connor et al.
Oncogene, 23(11), 2089-2095 (2004-03-17)
The identification of novel tumor-associated genes represents an important area of cancer research. To that end, we have discovered a number of genes whose expression is altered in breast tumors. One of these genes has been identified as the ring
Angelika Burger et al.
Neoplasia (New York, N.Y.), 8(8), 689-695 (2006-08-24)
Defects in ubiquitin E3 ligases are implicated in the pathogenesis of several human diseases, including cancer, because of their central role in the control of diverse signaling pathways. RING E3 ligases promote the ubiquitination of proteins that are essential to
Michael K Connor et al.
Molecular cancer research : MCR, 3(8), 453-461 (2005-08-27)
Ring finger proteins serve many vital functions within the cell. We have identified RNF11, a novel 154-amino acid ring finger-containing protein, which is elevated in breast cancer. Within its ring finger domain, RNF11 contains an AKT phosphorylation site (T135) that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service