Skip to Content
Merck
All Photos(1)

Key Documents

R0279

Sigma-Aldrich

Ro 23-9358

≥97% (HPLC), solid

Synonym(s):

N-[3,5-Bis(decyloxy)phenyl]-N-(carboxymethyl)glycine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$1,460.00

$1,460.00


Estimated to ship on28 May 2025



Select a Size

Change View
25 MG
$1,460.00

About This Item

Empirical Formula (Hill Notation):
C30H51NO6
CAS Number:
Molecular Weight:
521.73
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$1,460.00


Estimated to ship on28 May 2025


Assay

≥97% (HPLC)

form

solid

color

white

mp

99.5-110 °C

solubility

DMSO: 16 mg/mL

originator

Roche

storage temp.

2-8°C

SMILES string

CCCCCCCCCCOc1cc(OCCCCCCCCCC)cc(c1)N(CC(O)=O)CC(O)=O

InChI

1S/C30H51NO6/c1-3-5-7-9-11-13-15-17-19-36-27-21-26(31(24-29(32)33)25-30(34)35)22-28(23-27)37-20-18-16-14-12-10-8-6-4-2/h21-23H,3-20,24-25H2,1-2H3,(H,32,33)(H,34,35)

InChI key

SWOCSPJYINCHOW-UHFFFAOYSA-N

Biochem/physiol Actions

Diacylglycerol lipase inhibitor. Target for anti-obesity drugs.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service