Skip to Content
Merck
All Photos(1)

Key Documents

Q1878

Sigma-Aldrich

Quinine

Synonym(s):

6′-Methoxycinchonidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2
CAS Number:
Molecular Weight:
324.42
Beilstein:
91867
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

173-175 °C (lit.)

SMILES string

COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1

InChI

1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19-,20+/m0/s1

InChI key

LOUPRKONTZGTKE-WZBLMQSHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potassium channel blocker

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Azadmehr et al.
Bioelectrochemistry (Amsterdam, Netherlands), 127, 59-67 (2019-02-01)
This study employed a molecularly imprinted composite (MIC) to develop a selective and very sensitive sensor for the determination of quinine. To fabricate the MIC sensor, 3-methyl-4-nitrophenol (MNP) and L-tyrosine (Tyr) were simultaneously electrodeposited in acidic media containing HAuCl4 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service