Skip to Content
Merck
All Photos(1)

Documents

PZ0408

Sigma-Aldrich

WAY-163909 hydrochloride

≥98% (HPLC)

Synonym(s):

(7bR,10aR)-1,2,3,4,7a,9,10,10a-Octahydro-8H-cyclopenta[4,5]pyrrolo[3,2,1-jk][1,4]benzodiazepine, hydrochloride, (7bR,10aR)-1,2,3,4,8,9,10,10a-Octahydro-7bH-cyclopenta-[b][1,4]diazepino[6,7,1hi]indole, hydrochloride, WAY 163909 hydrochloride, WAY-162545 active isomer, WAY-909 hydrochloride, WAY163909 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2·HCl
CAS Number:
Molecular Weight:
250.77
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

room temp

InChI

1S/C14H18N2.ClH/c1-3-10-9-15-7-8-16-13-6-2-4-11(13)12(5-1)14(10)16;/h1,3,5,11,13,15H,2,4,6-9H2;1H/t11-,13-;/m1./s1

InChI key

UJVFYZZBCUXMDV-LOCPCMAASA-N

Biochem/physiol Actions

WAY-163909 is a 5-hydroxytryptamine receptor 2C (5-HT2C) agonist (EC50/Emax = 8 nM/90% vs. 185 nM/40%/5-HT2B; Ki = 10.5 nM/5-HT2C, 212 nM/5-HT2A, 343 nM/5-HT7, 485 nM/5-HT2B, 245 nM/D4, 665 nM/rat α-1, >1 μM/5-HT1A, D2, D3, rat 5-HT-T) with no 5-HT2A-stimulatory potency. WAY-163909 exhibits anorectic (ED50 in mg/kg via i.p. = 1.4/DIO mice and 5.19/Zucker rats), antipsychotic (morphine-induced mouse climbing ID50 = 10.69 mg/kg i.p.; rat avoidance responding reduction ED50 = 1.27 mg/kg i.p. and 6.50 mg/kg p.o.) and antidepressant (10 mg/kg, i.p. or s.c. in forced rat swimming test ; 3 mg/kg/day i.p. in reducing olfactory bulbectomy/BULB-induced hyperactivity in rats) efficacy in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen L Marquis et al.
The Journal of pharmacology and experimental therapeutics, 320(1), 486-496 (2006-10-14)
Serotonin-2C (5-HT2C) receptor antagonists and agonists have been shown to affect dopamine (DA) neurotransmission, with agonists selectively decreasing mesolimbic DA. As antipsychotic efficacy is proposed to be associated with decreased mesolimbic DA neurotransmission by virtue of DA D2 receptor antagonism
Maylen Perez Diaz et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 44(3), 478-486 (2018-09-07)
Perseverative behavior has been highly implicated in addiction. Activation of serotonin 2C receptors (5-HT2CRs) attenuates cocaine and high caloric food intake, but whether a 5-HT2CR agonist can reduce high caloric diet (HCD) or methamphetamine (METH) intake and response perseveration remains
Benjamin U Phillips et al.
Psychopharmacology, 235(7), 2101-2111 (2018-04-24)
Dysregulation of the serotonin (5-HT) system is a pathophysiological component in major depressive disorder (MDD), a condition closely associated with abnormal emotional responsivity to positive and negative feedback. However, the precise mechanism through which 5-HT tone biases feedback responsivity remains
John Dunlop et al.
The Journal of pharmacology and experimental therapeutics, 313(2), 862-869 (2005-02-12)
The pharmacological profile of WAY-163909 [(7bR, 10aR)-1,2,3,4,8,9,10,10a-octahydro-7bH-cyclopenta-[b][1,4]diazepino[6,7,1hi]indole], a novel 5-hydroxytryptamine (HT)(2C) (serotonin) receptor-selective agonist is presented. WAY-163909 displaced [(125)I]2,5-dimethoxy-4-iodoamphetamine binding from human 5-HT(2C) receptor sites, in Chinese hamster ovary (CHO) cell membranes, with a K(i) value of 10.5 +/- 1.1
Sharon Rosenzweig-Lipson et al.
Psychopharmacology, 192(2), 159-170 (2007-02-14)
Activation of one or more of the serotonin (5-HT) receptors may play a role in mediating the antidepressant effects of SSRIs. The present studies were conducted to evaluate the effects of the novel 5-HT2C receptor agonist WAY-163909 in animal models

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service