Skip to Content
Merck
All Photos(1)

Key Documents

PZ0365

Sigma-Aldrich

CP-376395 hydrochloride

≥98% (HPLC)

Synonym(s):

4-Pyridinamine, N-(1-ethylpropyl)-3,6-dimethyl-2-(2,4,6-trimethylphenoxy)-, hydrochloride (1:1), CP376395 hydrochloride, N-(1-Ethylpropyl)-3,6-dimethyl-2-(2,4,6-trimethylphenox y)-4-pyridinamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$127.00
25 MG
$512.00

$127.00


Check Cart for Availability
New, lower price on this item!


Select a Size

Change View
5 MG
$127.00
25 MG
$512.00

About This Item

Empirical Formula (Hill Notation):
C21H30N2O · HCl
CAS Number:
Molecular Weight:
362.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$127.00


Check Cart for Availability
New, lower price on this item!

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=C(NC(CC)CC)C=C(C)N=C1OC2=C(C)C=C(C)C=C2C.[H]Cl

InChI

1S/C21H30N2O.ClH/c1-8-18(9-2)23-19-12-16(6)22-21(17(19)7)24-20-14(4)10-13(3)11-15(20)5;/h10-12,18H,8-9H2,1-7H3,(H,22,23);1H

InChI key

UDIOZDHQQNYISB-UHFFFAOYSA-N

Biochem/physiol Actions

CP-376395 is a potent and selective antagonist of the CRF1 receptor, which has been shown to play an important role in mediating behavioral and endocrine responses to fear and stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey A Simms et al.
Addiction biology, 19(4), 606-611 (2013-02-01)
Corticotrophin-releasing factor (CRF) is a mediator of stress responses and a key modulator of ethanol-mediated behaviors. We report here that the CRF receptor 1 (CRF-R1) antagonist, CP-376395 reduces 20% ethanol consumption in animals trained to consume ethanol on an intermittent
Yuhpyng L Chen et al.
Journal of medicinal chemistry, 51(5), 1385-1392 (2008-02-22)
An orally active clinical candidate of corticotropin-releasing factor 1 (CRF 1) antagonist 1 showed a significant positive food effect in dog and human after oral administration. Efforts to address the food effect issue led us to explore and discover compounds
Andrew S Dore et al.
Current molecular pharmacology, 10(4), 334-344 (2017-02-12)
The structural analysis of class B G protein-coupled receptors (GPCR), cell surface proteins responding to peptide hormones, has until recently been restricted to the extracellular domain (ECD). Corticotropin-releasing factor receptor type 1 (CRF1R) is a class B receptor mediating stress

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service