Skip to Content
Merck
All Photos(1)

Key Documents

PZ0325

Sigma-Aldrich

PF-CBP1

≥98% (HPLC)

Synonym(s):

4-(2-(5-(3,5-Dimethylisoxazol-4-yl)-2-(4-propoxyphenethyl)-1H-benzo[d]imidazol-1-yl)ethyl)morpholine, 5-(Dimethyl-1,2-oxazol-4-yl)-1-[2-(morpholin-4-yl)ethyl]-2-[2-(4-propoxyphenyl)ethyl]-1H-1,3-benzodiazole, PF CBP1, PF-06670910

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00
25 MG
$503.00

$141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$141.00
25 MG
$503.00

About This Item

Empirical Formula (Hill Notation):
C29H36N4O3
Molecular Weight:
488.62
UNSPSC Code:
12352200
NACRES:
NA.77

$141.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

General description

The gene CBP (CREB binding protein) is mapped to human chromosome 16p13.[1]

Biochem/physiol Actions

CBP is a transcriptional coactivator. The protein is a link between the DNA-associated transcription factors and the RNA polymerase 2 complex. It also exhibits histone acetyltransferase activity.[2]
PF-CBP1 is potent and highly-selective inhibitor of the bromodomain of CREB binding protein (CBP BRD) that down regulates targets of CBP in macrophages primary neurons. Also, PF-CBP1 significantly reduces levels of RGS4 mRNA levels in neurons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kunka Kamenarova et al.
Human pathology, 47(1), 144-149 (2015-11-26)
Rubinstein-Taybi syndrome (RSTS) is a rare autosomal dominant congenital disorder (prevalence, 1:125000-720000) characterized by broad thumbs and halluces, facial dysmorphism, psychomotor development delay, skeletal defects, abnormalities in the posterior fossa, and short stature. The purpose of this study was to
Natalie H Theodoulou et al.
ChemMedChem, 11(5), 477-487 (2016-01-11)
The bromodomain and extra terminal (BET) family of bromodomains have been the focus of extensive research, leading to the development of many potent, selective chemical probes and recent clinical assets. The profound biology associated with BET bromodomain inhibition has provided
Spontaneous complete and sustained remission of a rearrangement CBP (16p13)-positive disseminated congenital myelosarcoma.
Carl Friedrich Classen et al.
Annals of hematology, 84(4), 274-275 (2004-12-18)
Eugene L Piatnitski Chekler et al.
Chemistry & biology, 22(12), 1588-1596 (2015-12-17)
Bromodomains are involved in transcriptional regulation through the recognition of acetyl lysine modifications on diverse proteins. Selective pharmacological modulators of bromodomains are lacking, although the largely hydrophobic nature of the pocket makes these modules attractive targets for small-molecule inhibitors. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service