Skip to Content
Merck
All Photos(1)

Key Documents

PZ0268

Sigma-Aldrich

AG-17724

≥98% (HPLC)

Synonym(s):

(R)-6-Fluoro--[(2-naphthalenylcarbonyl)amino]-1H-benzimidazole-2-propanoic acid, AG 17724

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$150.00
25 MG
$434.00

$150.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$150.00
25 MG
$434.00

About This Item

Empirical Formula (Hill Notation):
C21H16FN3O3
CAS Number:
Molecular Weight:
377.37
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$150.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

OC([C@H](NC(C1=CC(C=CC=C2)=C2C=C1)=O)CC3=NC4=C(C=CC(F)=C4)N3)=O

InChI

1S/C21H16FN3O3/c22-15-7-8-16-17(10-15)24-19(23-16)11-18(21(27)28)25-20(26)14-6-5-12-3-1-2-4-13(12)9-14/h1-10,18H,11H2,(H,23,24)(H,25,26)(H,27,28)/t18-/m1/s1

InChI key

NKMPZFCFXCJBEY-GOSISDBHSA-N

Biochem/physiol Actions

AG-17724 is a potent peptidyl-prolyl isomerase (PPIase) Pin1 inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service