Skip to Content
Merck
All Photos(1)

Key Documents

PZ0243

Sigma-Aldrich

PF-06672131

≥95% (HPLC)

Synonym(s):

(2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-(2-propyn-1-yloxy)-6-quinazolinyl]-4-(dimethylamino)-2-butenamide, (E)-N-(4-((3-Chloro-4-fluorophenyl)amino)-7-(prop-2-yn-1-yloxy)quinazolin-6-yl)-4-(dimethylamino)but-2-enamide, Probe 8

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$127.00
25 MG
$510.00

$127.00


Estimated to ship on30 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$127.00
25 MG
$510.00

About This Item

Empirical Formula (Hill Notation):
C23H21ClFN5O2
CAS Number:
Molecular Weight:
453.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$127.00


Estimated to ship on30 May 2025

New, lower price on this item!

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

FC(C=C1)=C(Cl)C=C1NC2=NC=NC3=CC(OCC#C)=C(NC(/C=C/CN(C)C)=O)C=C32

InChI

1S/C23H21ClFN5O2/c1-4-10-32-21-13-19-16(12-20(21)29-22(31)6-5-9-30(2)3)23(27-14-26-19)28-15-7-8-18(25)17(24)11-15/h1,5-8,11-14H,9-10H2,2-3H3,(H,29,31)(H,26,27,28)/b6-5+

InChI key

XZAHPCGWUZFHBI-AATRIKPKSA-N

Biochem/physiol Actions

PF-06672131 is a selective EGFR kinase inhibitor thought to covalently react with active-site cysteine residues in the ATP binding pocket. PF-06672131 has been used as a probe for proteomic analysis to identify off-target binding substrates. The dimethylaminomethyl (DMAM) group appears to increase compound stability in cancer cells with a resultant increase in protein reactivity from selective EGFR binding to widespread proteome-wide reactivity after treatment.
PF-06672131 is a selective EGFR kinase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service