Skip to Content
Merck
All Photos(1)

Key Documents

PZ0139

Sigma-Aldrich

SC-58125

≥98% (HPLC)

Synonym(s):

5-(4-Fluorophenyl)-1-[4-(methylsulfonyl)phenyl]-3-(trifluoromethyl)pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12N2SO2F4
CAS Number:
Molecular Weight:
384.35
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

CS(=O)(=O)c1ccc(cc1)-n2nc(cc2-c3ccc(F)cc3)C(F)(F)F

InChI

1S/C17H12F4N2O2S/c1-26(24,25)14-8-6-13(7-9-14)23-15(10-16(22-23)17(19,20)21)11-2-4-12(18)5-3-11/h2-10H,1H3

InChI key

JHBIMJKLBUMNAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

SC-58125 is a selective cyclooxygenase 2 (COX-2) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Bancos et al.
Cellular immunology, 258(1), 18-28 (2009-04-07)
The widely used non-steroidal anti-inflammatory drugs (NSAIDs) function mainly through inhibition of cyclooxygenases 1 and 2 (Cox-1 and Cox-2). Unlike Cox-1, Cox-2 is considered an inducible and pro-inflammatory enzyme. We previously reported that Cox-2 is upregulated in activated human B
Juan Ding et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 25(2), 202-205 (2005-08-25)
The inhibitory effects of two kinds of selective cyclooxygenase-2 inhibitors on the proliferation of human carcinoma of larynx Hep-2 in vitro and their corresponding mechanisms were investigated. Hep-2 cells were cultured with two kinds of selective cyclooxygenase-2 inhibitors (Sc-58125 and
Tomofumi Hoshino et al.
The Tohoku journal of experimental medicine, 216(1), 53-59 (2008-08-23)
Acoustic injury is a common cause of hearing loss for people in industrial societies. Cyclooxygenase (COX) and lipoxygenase (LOX) are two important enzymes involved in arachidonic acid metabolism. Two COX isozymes are characterized, COX-1 and COX-2, that differ in terms
Jie Ding et al.
Molecular carcinogenesis, 45(4), 250-259 (2005-12-31)
Considering possible tumorigenic activity of cyclooxygenase (COX) isozymes in myeloma, we examined expression levels of COX-1 and -2 in seven human myeloma cell lines (ARH-77, IM-9, RPMI-8226, HPC, HS-Sultan, TSPC-1, and U-266). As analyzed by reverse transcriptase-polymerase chain reaction (RT-PCR)
Jian Zhang et al.
Ai zheng = Aizheng = Chinese journal of cancer, 26(4), 377-381 (2007-04-14)
Cyclooxygenase-2 (COX-2) is related closely to the tumorigenesis of bladder cancer, and COX-2 inhibitor has potential antitumor effect. This study was to investigate the effects of selective COX-2 inhibitors on the proliferation and apoptosis of human bladder cancer cell line

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service