Skip to Content
Merck
All Photos(1)

Documents

PZ0125

Sigma-Aldrich

CP-53631

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18BrN · HCl
CAS Number:
Molecular Weight:
352.70
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CN[C@@H]1CCC(c2ccc(Br)cc2)c3ccccc13

InChI

1S/C17H18BrN.ClH/c1-19-17-11-10-14(12-6-8-13(18)9-7-12)15-4-2-3-5-16(15)17;/h2-9,14,17,19H,10-11H2,1H3;1H/t14-,17+;/m0./s1

InChI key

FRGRVQZKRCFTHR-SQQLFYIASA-N

Biochem/physiol Actions

CP-53631 is a selective serotonin reuptake inhibitor (SSRI); antidepressant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service