Skip to Content
Merck
All Photos(1)

Key Documents

PZ0104

Sigma-Aldrich

CP-91149

≥98% (HPLC)

Synonym(s):

5-Chloro-N-[(1S,2R)-3-(dimethylamino)-2-hydroxy-3-oxo-1-(phenylmethyl)propyl]-1H-indole-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$200.00
25 MG
$637.00

$200.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$200.00
25 MG
$637.00

About This Item

Empirical Formula (Hill Notation):
C21H22ClN3O3
CAS Number:
Molecular Weight:
399.87
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$200.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

CN(C)C(=O)[C@H](O)[C@H](Cc1ccccc1)NC(=O)c2cc3cc(Cl)ccc3[nH]2

InChI

1S/C21H22ClN3O3/c1-25(2)21(28)19(26)17(10-13-6-4-3-5-7-13)24-20(27)18-12-14-11-15(22)8-9-16(14)23-18/h3-9,11-12,17,19,23,26H,10H2,1-2H3,(H,24,27)/t17-,19+/m0/s1

InChI key

HINJNZFCMLSBCI-PKOBYXMFSA-N

Biochem/physiol Actions

CP-91149 is a selective glycogen phosphorylase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service