Skip to Content
Merck
All Photos(2)

Documents

PZ0012

Sigma-Aldrich

Sunitinib malate

≥98% (HPLC), powder, receptor tyrosine kinase inhibitor

Synonym(s):

N-[2-(Diethylamino)ethyl]-5-[(Z)-(5-fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxamide (2S)-2-hydroxybutanedioic acid (1:1) salt, SU 011248, SU 11248, SU112248

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27FN4O2 · C4H6O5
CAS Number:
Molecular Weight:
532.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Sunitinib malate, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

O[C@@H](CC(O)=O)C(O)=O.CCN(CC)CCNC(=O)c1c(C)[nH]c(\C=C2/C(=O)Nc3ccc(F)cc23)c1C

InChI

1S/C22H27FN4O2.C4H6O5/c1-5-27(6-2)10-9-24-22(29)20-13(3)19(25-14(20)4)12-17-16-11-15(23)7-8-18(16)26-21(17)28;5-2(4(8)9)1-3(6)7/h7-8,11-12,25H,5-6,9-10H2,1-4H3,(H,24,29)(H,26,28);2,5H,1H2,(H,6,7)(H,8,9)/b17-12-;/t;2-/m.0/s1

InChI key

LBWFXVZLPYTWQI-IPOVEDGCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sunitinib malate has been used:
  • to study its mechanism in the inhibition of MCF-7 cell migration and angiogenesis
  • as a reference molecule to compare its docking energies with other ligands/vascular endothelial growth factor receptor (VEGF) receptor blocker
  • used in proliferation assay

Biochem/physiol Actions

Sunitinib has greater bioavailability and potency compared to other inhibitors. It prevents angiogenesis.
Sunitinib malate is a receptor tyrosine kinase inhibitor, which targets VEGF-R1, VEGF-R2, VEGF-R3, PDGF-Rα, PDGF-Rβ, KIT, FLT3, CSF-1R, and RET. Sunitinib malate is an anticancer drug.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iris Uribesalgo et al.
EMBO molecular medicine, 11(8), e9266-e9266 (2019-07-04)
Angiogenesis is a hallmark of cancer, promoting growth and metastasis. Anti-angiogenic treatment has limited efficacy due to therapy-induced blood vessel alterations, often followed by local hypoxia, tumor adaptation, progression, and metastasis. It is therefore paramount to overcome therapy-induced resistance. We
Sunitinib: from rational design to clinical efficacy
Chow LQM and Eckhardt SG
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 25(7), 884-896 (2007)
Nils Ohnesorge et al.
Frontiers in pharmacology, 10, 508-508 (2019-06-11)
Unbiased screening of large randomized chemical libraries in vivo is a powerful tool to find new drugs and targets. However, forward chemical screens in zebrafish can be time consuming and usually >99% of test compounds have no significant effect on
Kingkamon Junkunlo et al.
Stem cells and development, 26(20), 1449-1459 (2017-08-15)
The platelet-derived growth factor (PDGF) receptor, a tyrosine kinase (TK) receptor whose ligand is PDGF, is crucial in the transduction of extracellular signals into cells and mediates numerous processes, such as cell proliferation, differentiation, survival, and migration. We demonstrate the
In-silico screening and In-vitro validation of Vascular Endothelial Growth Factor Receptor-2 (VEGFR-2) inhibitors
Saraswat D, et al.
Bioinformation, 10(5), 273-273 (2014)

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service