Skip to Content
Merck
All Photos(1)

Key Documents

PLA0287

Sigma-Aldrich

Rabbit anti-CDK1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CDC2, CDC28A, CDK1, G1 to S and G2 to M, P34CDC2, cell cycle controller CDC2, cell division control protein 2 homolog, cell division cycle 2, cell division protein kinase 1, p34 protein kinase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$512.00

$512.00


Estimated to ship on13 May 2025



Select a Size

Change View
100 μL
$512.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$512.00


Estimated to ship on13 May 2025


biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 2- 10 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_001777.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... CDK1(983)

Immunogen

The epitope recognized by PLA0287 maps to a region between residue 1 and 50 of human Cyclin-Dependent Kinase 1 using the numbering given in entry NP_001777.1 (GeneID 983).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Cyclin-dependent kinase 1 (CDK1) is a member of the Ser/Thr protein kinase family. CDK1 is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF), which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Mitotic cyclins stably associate with this protein and function as regulatory subunits. The kinase activity of CKD1 is controlled by cyclin accumulation and destruction through the cell cycle. The phosphorylation and dephosphorylation of this protein also play important regulatory roles in cell cycle control [taken from NCBI Entrez Gene (Gene ID: 983)].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Zhao et al.
Oncology letters, 17(1), 42-54 (2019-01-19)
The present study examined the radiation biological response of cancer cells to different fractional irradiation doses and investigates the optimal fractional irradiation dose with improved biological effects. Radiobiological studies were performed at the molecular and cellular levels to provide insights

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service