PLA0224
Rabbit anti-IKK-alpha Antibody, Affinity Purified
Powered by Bethyl Laboratories, Inc.
Synonym(s):
Conserved helix-loop-helix ubiquitous kinase, I-kappa-B kinase 1, I-kappa-B kinase alpha, I-kappa-B kinase-alpha, IKK-A, IKK-a kinase, IKK-alpha, IKK1, IKKA, IkB kinase alpha subunit, IkBKA, IkappaB kinase, NFKBIKA, Nuclear factor NF-kappa-B inhibitor kinase alpha, Nuclear factor NFkappaB inhibitor kinase alpha, TCF-16, TCF16, transcription factor 16
Select a Size
$602.00
Estimated to ship on24 May 2025
Select a Size
About This Item
$602.00
Estimated to ship on24 May 2025
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
grade
Powered by Bethyl Laboratories, Inc.
species reactivity
human, mouse
technique(s)
immunohistochemistry: 1:100- 1:500
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000
accession no.
NP_001269.3
shipped in
wet ice
storage temp.
2-8°C
Gene Information
rabbit ... IKK-alpha(1147)
Immunogen
Physical form
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service