Skip to Content
Merck
All Photos(2)

Key Documents

PLA0074

Sigma-Aldrich

Rabbit anti-BARD1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

BARD-1, BRCA1-associated RING domain gene 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$602.00

$602.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$602.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$602.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:500-1:2,500

accession no.

NP_000456.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... BARD1(580)

Immunogen

The epitope recognized by PLA0074 maps to a region between residues 250 and 300 of human BRCA1-associated RING Domain 1 using the numbering given in entry NP_000456.1 (GeneID 580).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

BRCA1 associated RING domain 1 (BARD1) together with BRCA1 is an E3 ubiquitin ligase. BARD1/BRCA1 has been shown to ubiquinate RNA polymerase II in response to DNA damage. BARD1/BRCA1 ubiquitin ligase activity has also been shown to be important in the prevention of double stranded breaks and the regulation of centrosome number.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service