Skip to Content
Merck
All Photos(2)

Key Documents

PLA0017

Sigma-Aldrich

Rabbit anti-SMC1 Antibody, Affinity Purified

Synonym(s):

CDLS2, DXS423E, KIAA0178, SB1.8, SMC protein 1A, SMC-1-alpha, SMC-1A, SMC1, SMC1 (structural maintenance of chromosomes 1; yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1 (yeast), SMC1L1, SMC1alpha, Smcb, segregation of mitotic chromosomes 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$512.00

$512.00


Check Cart for Availability


Select a Size

Change View
100 μL
$512.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$512.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:2,000-1:10,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_006297.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... SMC1(8243)

Immunogen

The epitope recognized by PLA0017 maps to a region between residue 1175 and the C-terminus of human Structural Maintenance of Chromosomes 1 using the numbering given in entry NP_006297.2 (GeneID 8243).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Structural maintenance of chromosomes (SMC) proteins are part of the cohesin and condensin multisubunit complexes that play a critical role in chromosome organization, segregation, gene regulation, and DNA repair. The core cohesin complex includes a heterodimer of SMC1 and SMC3 that functions by forming a ring-like structure that holds sister chromatids together. Cohesin is important to sister chromatid separation and segregation during mitosis and meiosis and also plays a role in double-strand break repair by homologous recombination.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service