Skip to Content
Merck
All Photos(3)

Key Documents

PLA0009

Sigma-Aldrich

Rabbit anti-USP7 Antibody, Affinity Purified

Synonym(s):

HAUSP, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, TEF1, deubiquitinating enzyme 7, ubiquitin specific protease 7 (herpes virus-associated), ubiquitin thioesterase 7, ubiquitin-specific-processing protease 7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$512.00

$512.00


Check Cart for Availability


Select a Size

Change View
100 μL
$512.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$512.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:500- 1:2,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_003461.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... USP7(7874)

Immunogen

The epitope recognized by PLA0009 maps to a region between residues 1 and 50 of human Ubiquitin Specific Peptidase 7 using the numbering given in entry NP_003461.1 (GeneID 7874).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Ubiquitin (Ub) is a highly conserved protein found ubiquitously in eukaryotic organisms. The conjugation of ubiquitin to proteins is an important means to regulate protein activity for many cellular processes by tagging them for degradation. Removal of Ub can rescue proteins from degradation. This is accomplished by the ubiquitin-specific processing protease (UBP) family of enzymes. Ubiquitin specific processing protease 7 (USP7) is a member of this family that has been shown to interact and stabilize p53.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santiago Vidal et al.
Journal of virology, 94(1) (2019-10-11)
Some viruses take advantage of conjugation of ubiquitin or ubiquitin-like proteins to enhance their own replication. One example is Ebola virus, which has evolved strategies to utilize these modification pathways to regulate the viral proteins VP40 and VP35 and to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service