Skip to Content
Merck
All Photos(1)

Key Documents

P9792

Sigma-Aldrich

Phytase from Aspergillus ficuum

lyophilized powder, 1-5 units/mg solid (as phosphatase activity)

Synonym(s):

3-Phytase, myo-Inositol-hexakisphosphate 3-phosphohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

1-5 units/mg solid (as phosphatase activity)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Quality

May contain glucosidases and phosphatases.

Unit Definition

One unit will liberate 1.0 μmole of inorganic P from 4.2 × 10-2 M Mg·K phytate per min at pH 2.5 at 37 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G C Irving et al.
Journal of bacteriology, 112(1), 434-438 (1972-10-01)
The fungus Aspergillus ficuum NRRL 3135 is known to produce an extracellular nonspecific orthophosphoric monoester phosphohydrolase (EC 3.1.3.2) with a pH optimum of 2.0, as well as an extracellular myo-inositol hexaphosphate phosphohydrolase (EC 3.1.3.8; phytase) with pH optima of 2.0
L Jermutus et al.
Journal of biotechnology, 85(1), 15-24 (2001-02-13)
Thermostability is a key feature for commercially attractive variants of the fungal enzyme phytase. In an initial set of experiments, we restored ionic interactions and hydrogen bonds on the surface of Aspergillus terreus phytase, which are present in the homologous
Ion-exchange chromatography of inositol polyphosphates.
D J Cosgrove
Annals of the New York Academy of Sciences, 165(2), 677-686 (1969-10-17)
Nada M Tamim et al.
Journal of agricultural and food chemistry, 51(16), 4687-4693 (2004-01-07)
Phytate phosphorus (PP) hydrolysis by a 3-phytase was studied in vitro at pH 2.5 and 6.5 with either 0, 1.0, 4.0, or 9.0 g of Ca/kg diet, or 0, 1.0, 5.0, 7.5, or 10.0 g/kg diet of micro-mineral premix added
Duane F Berry et al.
Bioorganic & medicinal chemistry letters, 15(12), 3157-3161 (2005-05-10)
A novel approach for measuring phytase activity is presented. We have developed a new chromophoric substrate analog of phytic acid, 5-O-[6-(benzoylamino)hexyl]-d-myo-inositol-1,2,3,4,6-pentakisphosphate that permits direct measurement of the phosphate ester bond-cleavage reaction using HPLC. This compound, along with its dephosphorylated T-phosphatidylinositol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service