Skip to Content
Merck
All Photos(1)

Key Documents

P8898

Sigma-Aldrich

1,2-Diundecanoyl-sn-glycero-3-phosphocholine

~99%

Synonym(s):

L-α-Phosphatidylcholine, diundecanoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H60NO8P
CAS Number:
Molecular Weight:
593.77
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

functional group

phospholipid

shipped in

ambient

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCC)=O)COC(CCCCCCCCCC)=O)=O

InChI

1S/C30H61NO8P/c1-6-8-10-12-14-16-18-20-22-29(32)36-26-28(27-38-40(34,35)37-25-24-31(3,4)5)39-30(33)23-21-19-17-15-13-11-9-7-2/h28H,6-27H2,1-5H3,(H,34,35)

InChI key

VQTBCFRFEYMMQC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis Raj Gandhi Amrit et al.
PLoS genetics, 12(2), e1005788-e1005788 (2016-02-11)
Elimination of the proliferating germline extends lifespan in C. elegans. This phenomenon provides a unique platform to understand how complex metazoans retain metabolic homeostasis when challenged with major physiological perturbations. Here, we demonstrate that two conserved transcription regulators essential for
Chih-Chun Janet Lin et al.
Nature cell biology, 19(5), 550-557 (2017-04-25)
Microorganisms and their hosts share the same environment, and microbial metabolic molecules (metabolites) exert crucial effects on host physiology. Environmental factors not only shape the composition of the host's resident microorganisms, but also modulate their metabolism. However, the exact molecular
Ramesh Ratnappan et al.
PLoS genetics, 10(12), e1004829-e1004829 (2014-12-05)
In C. elegans, removal of the germline extends lifespan significantly. We demonstrate that the nuclear hormone receptor, NHR-49, enables the response to this physiological change by increasing the expression of genes involved in mitochondrial β-oxidation and fatty-acid desaturation. The coordinated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service