This product does not include tips. The manufacturer recommends polycarbonate tips with an OD of 0.34mm (340 µm) for use with this instrument. The volume range for each delivery is between 0.25µL and 3µL. The ID is not specified.
P7289
Drummond Sequencing Pipette
volume 0.05-3 μL , adjustable in 0.05 μL increment
Select a Size
$826.00
Estimated to ship on01 June 2025
Select a Size
About This Item
$826.00
Estimated to ship on01 June 2025
Recommended Products
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Drummond Scientific 3-000-203
volume
0.05-3 μL , adjustable in 0.05 μL increment
Related Categories
General description
Includes 6 tips (0.34 mm O.D.), two replacement O-rings, and one plastic spacer.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
What is the ID and OD of the tip? what is the material of the tip? Is the tip a tapped shape to avoid clog? I am transferring suspension liquid with particle size at 25um blended with water.
1 answer-
Helpful?
-