P6361
Protease S from Pyrococcus furiosus
recombinant, expressed in Bacillus sp.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in Bacillus sp.
form
solution
mol wt
45 kDa
concentration
100 units/mL
shipped in
wet ice
storage temp.
2-8°C
Application
Protease S is from Pyrococcus furiosus and is a recombinant protease that is expressed in Bacillus sp.. It is used for fragmentation of proteins and peptides required for primary structure analysis.
Biochem/physiol Actions
Protease S is a serine endoprotease with broad specificity that will digest native and denatured proteins. Cleavage occurs mainly on the carboxy side of peptide bonds. The optimal temperature range is 85 to 95 °C and the optimal pH range is 6.0 to 8.0. Protease S is inhibited by PMSF
Thermostable serine protease with broad specificity for native and denatured proteins.
Unit Definition
One unit will hydrolyze 1 μmol of N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide per minute at pH 7.0 at 95 °C.
Physical form
Solution in 25 mM Tris-HCl, pH 7.6, containing 40% ethanol.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
78.8 °F
Flash Point(C)
26 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service