Skip to Content
Merck
All Photos(1)

Key Documents

P6101

Sigma-Aldrich

Anti-Platelet-Derived Growth Factor antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-PDGF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
Neutral
WB
clone:
polyclonal
species reactivity:
human
citations:
10
technique(s):
indirect ELISA: suitable
neutralization: suitable
western blot: suitable

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Platelet-derived growth factor (PDGF) is a serum growth factor purified from human platelets. The members of PDGF are homodimers of A-, B-, C- and –D polypeptide chains and an AB heterodimer. PDGF is a mitogen and stimulates the proliferation of cells of connective tissue and glial cells, survival, differentiation, chemotaxis and wound healing. The cellular effects of PDGF isoforms are mediated by binding to α- and β-receptors which leads to activation of downstream signaling via PI3K, STAT and Ras/MAPK. Overexpression of PDGF isoforms has been observed in various malignancies including gliomas, osetosarcoma, chronic myelomonocytic leukemia and non-small-cell lung cancers
Anti-Human Platelet-Derived Growth Factor (PDGF) specifically recognizes natural human PDGF. It will also neutralize the activity of recombinant human PDGF-AB, recombinant human PDGF-AA, recombinant human PDGF-BB, and natural porcine PDGF-BB. It shows cross reactivity with recombinant human PDGF-AB, recombinant human PDGF-AA, recombinant human PDGF-BB, and natural porcine PDGF-BB.

Specificity

The antibody will neutralize the biological activity of natural human PDGF, recombinant human PDGF-AB, PDGF-AA and PDGF-BB and natural porcine PDGF-BB.

Immunogen

natural human PDGF.

Application

Anti-Human Platelet-Derived Growth Factor (PDGF) antibody may be used for ELISA at a working concentration of 0.25 μg/ml. The antibody has been used for ELISA at a dilution of 1:500 using human serum.. For immunoblotting, a concentration of 1 μg/ml is recommended. The antibody is suitable for neutralization reactions and has been used to neutralize PDGF of primary Human dermal fibroblasts (HDFs) and human platelet-enriched plasma.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline (pH 7.4) with 5% trehalose.

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanna Lindberg et al.
Upsala journal of medical sciences, 117(2), 92-98 (2012-03-02)
Platelet-derived growth factor B (PDGF-B) is a growth factor promoting and regulating cell migration, proliferation, and differentiation, involved in both developmental processes and in maintaining tissue homeostasis under strict regulation. What are the implications of prolonged or uncontrolled growth factor
Mechanism of human dermal fibroblast migration driven by type I collagen and Platelet-derived Growth Factor-BB
Li W et al
Molecular and Cellular Biology, 15, 294-309 (2004)
David J Margolis et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 17(10), 1822-1829 (2009-07-30)
Venous leg ulcers are a prevalent nonhealing wound of the lower extremity. Although topically applied growth factors successfully improve wound repair in animal studies, similar studies on humans with venous leg ulcers have not been successful. This study was designed
PDGF and PDGF receptors in glioma
Nazarenko I et al
Upsala Journal of Medical Sciences, 112, 99-112 (2012)
Fabio Celotti et al.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society, 14(2), 195-202 (2006-04-25)
Platelet-enriched plasma (PRP) is used in therapy as a source of growth factors in bone fracture and wound healing; however, few data exist on its role in the different aspects of the healing process. The effect of PRP and of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service