Skip to Content
Merck
All Photos(1)

Key Documents

P5976

Sigma-Aldrich

Anti-Platelet-Derived Growth Factor-BB antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-PDGF-BB

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
$827.00

$827.00


Estimated to ship on02 June 2025



Select a Size

Change View
0.1 MG
$827.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

$827.00


Estimated to ship on02 June 2025


biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDGFB(5155)

General description

Platelet-Derived Growth Factor-BB (PDGF-BB) is a cystine-linked homodimer that belongs to PDGF growth factor family and is expressed mainly by vascular endothelium, megakaryocytes and Leydig cells. It has key roles in regulating neurogenesis, producing proinflammatory cytokines by orbital fibroblasts in Graves′ Ophthalmopathy, cell proliferation, development, angiogenesis and cell survival. Anti-Platelet-Derived Growth Factor-BB antibody can be used for neutralizing the biological activity of recombinant human PDGF-BB, recombinant human-AB, natural human PDGF, rat PDGF-BB, and porcine PDGF-BB. It can also neutralize recombinant human PDGF-AA at 50-100 fold high IgG concentration. Goat Anti-Platelet-Derived Growth Factor-BB antibody reacts specifically with human PDGF-BB. The product has also shown less than 10% cross-reactivity for recombinant human PDGF-AA.

Immunogen

Recombinant human PDGF-BB expressed in Escherichia coli.

Application

Anti-Platelet-Derived Growth Factor-BB antibody can be used in immunoblotting and ELISA.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline pH 7.4 with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leendert van Steensel et al.
Investigative ophthalmology & visual science, 51(2), 1002-1007 (2009-10-03)
Graves' ophthalmopathy (GO) is characterized by the infiltration of immune cells into the orbit, a process in which cytokines play a central role. Orbital fibroblasts are potent producers of cytokines on different stimuli. Recently, the authors showed increased expression of
Honghong Yao et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(29), 9835-9847 (2012-07-21)
Platelet-derived growth factor-BB (PDGF-BB) has been reported to provide tropic support for neurons in the CNS. However, whether PDGF-BB regulates neurogenesis, especially in the context of HIV-associated neurological disorder and drug abuse, remains essentially unknown. In this study, we demonstrate
Renée V Hoch et al.
Development (Cambridge, England), 130(20), 4769-4784 (2003-09-04)
Recent advances in genetic manipulation have greatly expanded our understanding of cellular responses to platelet-derived growth factors (PDGFs) during animal development. In addition to driving mesenchymal proliferation, PDGFs have been shown to direct the migration, differentiation and function of a
Johanna Andrae et al.
Genes & development, 22(10), 1276-1312 (2008-05-17)
Platelet-derived growth factors (PDGFs) and their receptors (PDGFRs) have served as prototypes for growth factor and receptor tyrosine kinase function for more than 25 years. Studies of PDGFs and PDGFRs in animal development have revealed roles for PDGFR-alpha signaling in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service