Skip to Content
Merck
All Photos(1)

Key Documents

P5963

Sigma-Aldrich

Platelet-derived Growth Factor β−Receptor Fragment 742-758

≥95% (HPLC)

Synonym(s):

Asp-Met-Ser-Lys-Asp-Glu-Ser-Val-Asp-Tyr-Val-Pro-Met-Leu-Asp-Met-Lys

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$424.00

$424.00


Estimated to ship on28 May 2025



Select a Size

Change View
0.5 MG
$424.00

About This Item

Empirical Formula (Hill Notation):
C84H135N19O31S3
CAS Number:
Molecular Weight:
2003.27
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

$424.00


Estimated to ship on28 May 2025


Assay

≥95% (HPLC)

form

powder

storage temp.

−20°C

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service