Skip to Content
Merck
All Photos(1)

Key Documents

P4775

Sigma-Aldrich

1-Phenyl-1,3,8-triazaspiro[4.5]decan-4-one

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N3O
CAS Number:
Molecular Weight:
231.29
Beilstein:
617685
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥90%

storage temp.

2-8°C

SMILES string

O=C1NCN(c2ccccc2)C13CCNCC3

InChI

1S/C13H17N3O/c17-12-13(6-8-14-9-7-13)16(10-15-12)11-4-2-1-3-5-11/h1-5,14H,6-10H2,(H,15,17)

InChI key

HTQWGIHCFPWKAS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service