Skip to Content
Merck
All Photos(1)

Key Documents

P4429

Sigma-Aldrich

Pancreozymin from porcine intestine

2-6 Crick units/mg solid

Synonym(s):

CCK, Cholecystokinin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

solid

specific activity

2-6 Crick units/mg solid

foreign activity

secretin 0.4 - 1.0 Crick units/mg solid

storage temp.

2-8°C

InChI

1S/C166H261N51O52S4/c1-15-84(9)130(159(261)211-116(78-220)154(256)206-110(68-125(227)228)150(252)191-97(38-27-54-182-165(176)177)139(241)205-112(70-127(231)232)152(254)215-132(88-42-44-91(45-43-88)269-273(266,267)268)161(263)197-100(48-58-270-12)135(237)185-75-124(226)190-106(64-89-72-184-94-35-21-20-33-92(89)94)146(248)195-101(49-59-271-13)141(243)204-111(69-126(229)230)151(253)198-103(133(173)235)63-87-31-18-17-19-32-87)213-143(245)98(39-28-55-183-166(178)179)192-147(249)107(65-90-73-180-80-187-90)201-153(255)115(77-219)210-157(259)119-41-30-57-217(119)163(265)113(71-128(233)234)207-145(247)105(62-82(5)6)200-149(251)109(67-122(172)224)203-140(242)99(46-47-120(170)222)193-144(246)104(61-81(3)4)199-148(250)108(66-121(171)223)202-138(240)96(36-23-25-52-168)196-158(260)129(83(7)8)212-160(262)131(85(10)16-2)214-155(257)117(79-221)208-142(244)102(50-60-272-14)194-137(239)95(37-26-53-181-164(174)175)189-123(225)74-186-136(238)114(76-218)209-156(258)118-40-29-56-216(118)162(264)86(11)188-134(236)93(169)34-22-24-51-167/h17-21,31-33,35,42-45,72-73,80-86,93,95-119,129-132,184,218-221H,15-16,22-30,34,36-41,46-71,74-79,167-169H2,1-14H3,(H2,170,222)(H2,171,223)(H2,172,224)(H2,173,235)(H,180,187)(H,185,237)(H,186,238)(H,188,236)(H,189,225)(H,190,226)(H,191,252)(H,192,249)(H,193,246)(H,194,239)(H,195,248)(H,196,260)(H,197,263)(H,198,253)(H,199,250)(H,200,251)(H,201,255)(H,202,240)(H,203,242)(H,204,243)(H,205,241)(H,206,256)(H,207,247)(H,208,244)(H,209,258)(H,210,259)(H,211,261)(H,212,262)(H,213,245)(H,214,257)(H,215,254)(H,227,228)(H,229,230)(H,231,232)(H,233,234)(H4,174,175,181)(H4,176,177,182)(H4,178,179,183)(H,266,267,268)

InChI key

QFLBZJPOIZFFJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cholecystokinin (CCK) is a peptide hormone of the gastrointestinal system responsible for stimulating the digestion of fat and protein. It is used to study brain, kidney and pancreatic functioning as well as reproductive behavior and glucose tolerance[1][2].

Biochem/physiol Actions

CCK mediates digestion and satiety. It has anti-inflammatory properties. CCK stimulates insulin secretion through phospholipase activity in b-cells[2].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alicia Wall et al.
General and comparative endocrinology, 183, 44-52 (2013-01-12)
The effects of fasting and feeding on the brain expression of orexin (OX), tyrosine hydroxylase (TH), peptide Y (PY) and cholecystokinin (CCK) were examined in the blind cavefish Astyanax fasciatus mexicanus. A 10-days fasting period induced increases in both OX
Anita Belza et al.
The American journal of clinical nutrition, 97(5), 980-989 (2013-03-08)
Effects of protein intake on appetite-regulating hormones and their dynamics are unclear. We investigated the satiating effects of meals with varying protein contents and whether there was an effect of dose on appetite-regulating hormones and appetite ratings. Twenty-five men [mean
A conversation with Jeffrey M. Friedman by Ushma S. Neill.
Jeffrey M Friedman
The Journal of clinical investigation, 123(2), 529-530 (2013-04-20)
Alan R Mackie et al.
American journal of physiology. Gastrointestinal and liver physiology, 304(11), G1038-G1043 (2013-04-13)
The aim of this study was to determine the extent to which gastric layering and retention of a meal could be used to reduce appetite using the same caloric load. Liquid (control) and semi-solid (active) meals were produced with the
Utilization of cholecystokinin cholescintigraphy in clinical practice.
Bryan K Richmond et al.
Journal of the American College of Surgeons, 217(2), 317-323 (2013-06-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service