P4300
Pectinase from Rhizopus sp.
suitable for plant cell culture, BioReagent, crude powder
Synonym(s):
Macerozyme R-10, Poly-(1,4-α-D-galacturonide) glycanohydrolase, Polygalacturonase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product line
BioReagent
form
crude powder
specific activity
400-800 units/g solid
technique(s)
cell culture | plant: suitable
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Pectinase P4300 is a natural pectinase (polygalacturonase) derived from fungus Rhizops sp. that randomly hydrolyzes the (1→4)-α-D-galactosiduronic linkages in pectate and other galacturonans. This enzyme breaks down pectin found in the middle lamella of plant cell walls. Pectinase helps facilitate the maceration of plant materials and extraction processes. Pectinase P4300 is used in plant protoplast preparation to digest cell wall prior to organelle isolation.
Application
Used in plant protoplast preparation to digest cell wall prior to organelle isolation.
Unit Definition
One unit will liberate 1.0 μmole of galacturonic acid from polygalacturonic acid per min at pH 4.0 at 25 °C.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jeffrey A Mertens et al.
Fungal genetics and biology : FG & B, 45(12), 1616-1624 (2008-10-22)
A search of the recently sequenced Rhizopus oryzae strain 99-880 genome database uncovered 18 putative polygalacturonase genes with two genes being identical and only one with similarity to a previously reported R. oryzae polygalacturonase gene. The 17 different genes share
Shigenobu Yoshida et al.
Mycological research, 108(Pt 12), 1407-1414 (2005-03-11)
The polygalacturonase (PG)-encoding gene (rpg1) of Rhizopus oryzae, the causal pathogen of rhizopus rot of mulberry, was cloned and sequenced. PGs were partially purified from incubation mixture of 2% pectin medium and their N-terminal amino acid sequences were determined by
K Saito et al.
Journal of industrial microbiology & biotechnology, 30(7), 440-444 (2003-07-05)
Rhizopus oryzae strain NBRC 4707 produced lactic acid and ethanol more efficiently than strain NRRL 395 in potato pulp, an agricultural by-product of the starch industry. The two strains developed comparable activities of xylanase, cellulase, alpha-amylase, and glucoamylase, while the
Katsuichi Saito et al.
Microbiological research, 159(1), 83-86 (2004-05-27)
The pectinolytic enzyme from the solid-state culture of Rhizopus oryzae NBRC 4707 was purified to homogeneity by column chromatography on CM-Toyopearl 650 M and hydroxylapatite. The molecular weight of the enzyme was estimated by SDS-polyacrylamide gel electrophoresis to be 31,000
S S Ahmed et al.
Genetics and molecular research : GMR, 11(3), 3091-3104 (2012-09-26)
Molecular chaperones of plasmid pBI121 carrying CaMV35S promoter and a nucleotide sequence of plasmid pBI221 were inserted into plasmid pCAMBIA2300 to construct an intermediate vector: pVBG2307. This novel vector pVBG2307 contains a greatly expanded multiple cloning site with an adjacent
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service