Skip to Content
Merck
All Photos(1)

Key Documents

P4238

Sigma-Aldrich

Prion Protein Fragment 106-126 Scrambled Human

≥85% (HPLC), solid

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$530.00

$530.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.5 MG
$530.00

About This Item

Empirical Formula (Hill Notation):
C80H138N26O24S2
CAS Number:
Molecular Weight:
1912.24
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

$530.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥85% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PRNP(5621)

Amino Acid Sequence

Asn-Gly-Ala-Lys-Ala-Leu-Met-Gly-Gly-His-Gly-Ala-Thr-Lys-Val-Met-Val-Gly-Ala-Ala-Ala

Application

Control used in the study of the biological activity of prion protein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service