Skip to Content
Merck
All Photos(1)

Documents

P4119

Sigma-Aldrich

Polymyxin B sulfate salt

Biotechnology Performance Certified

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H96N16O13 · 2H2SO4
CAS Number:
Molecular Weight:
1385.61
EC Number:
MDL number:
UNSPSC Code:
51101500

grade

Biotechnology Performance Certified

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

solubility

H2O: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

O=C(C(NC(C(CCN)NC(CCCCC(C)CC)=O)=O)C(C)O)NC(CCN)C(NC(CCNC(C(NC(C(NC(C(CCN)N1)=O)CCN)=O)C(O)C)=O)C(NC(CCN)C(NC(CC2=CC=CC=C2)C(NC(CC(C)C)C1=O)=O)=O)=O)=O.O=S(O)(O)=O

InChI

1S/C48H82N16O13.H2O4S/c1-27(2)24-37-47(76)59-32(11-19-52)41(70)56-31(10-18-51)43(72)61-35(14-22-65)39(68)54-21-13-34(45(74)57-33(12-20-53)44(73)64-38(48(77)63-37)25-28-6-4-3-5-7-28)60-42(71)30(9-17-50)58-46(75)36(15-23-66)62-40(69)29(8-16-49)55-26-67;1-5(2,3)4/h3-7,26-27,29-38,65-66H,8-25,49-53H2,1-2H3,(H,54,68)(H,55,67)(H,56,70)(H,57,74)(H,58,75)(H,59,76)(H,60,71)(H,61,72)(H,62,69)(H,63,77)(H,64,73);(H2,1,2,3,4)

InChI key

HNDFYNOVSOOGDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Polymyxin B sulfate is a strongly cationic cyclic polypeptide antibiotic that is derived from fermentation of Bacilus polymyxa. It is a mixture of B1 and B2 sulfate. The product has been used clinically to treat infections of the urinary tract, meninges and blood stream caused by susceptible strains of Pseudomonas aeruginosa . It also has uses studying multidrug-resistant pathogens, as an immobilized agent for removal of endotoxins, and to induce pore formation in the membranes of cortex cells from excised sorghum roots.

Biochem/physiol Actions

Mode of Action: Polymyxin B Sulfate binds to the lipid A portion of bacterial lipopolysaccharides†, disrupting the cytoplasmic membrane by inducing pores large enough to permit nucleotide leakage in bacterial walls. This disrupts the permeability of the cytoplasmic membrane.

Antimicrobial spectrum: Has bactericidal action on most gram-negative bacilli††, including E. Coli and on most fungi and gram-positive bacteria.

Caution

As supplied, the product should be stored at 2-8°C in the dark. No change was observed in retained samples after three years′ of storage in these conditions. Stock solutions should be sterile filtered and stored at 2-8°C. They are stable at 37°C for 5 days.

Preparation Note

Polymyxin B sulfate is soluble in water at 50 mg/mL, producing a very slightly hazy, colorless to yellow solution. It has only minimal solubility in any organic solvent. For example, it has a solubility of 0.115 mg/mL in ethanol. This product is biotechnology performance certified.

Other Notes

10mu,25mu,50mu
Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Andrea Giacometti et al.
The Journal of antimicrobial chemotherapy, 49(1), 193-196 (2001-12-26)
The efficacy of two polymyxin-like peptides, KFFKFFKFF and IKFLKFLKFL, alone and combined with levofloxacin, was investigated in a rat model of septic shock. Rats were given an ip injection of 2 x 10(10) cfu of Escherichia coli and randomized to
Michael Hogardt et al.
The Journal of antimicrobial chemotherapy, 54(6), 1057-1061 (2004-10-29)
With their potent activity against Gram-negative bacteria, the polymyxins are important alternative antibiotics for cystic fibrosis (CF) patients. A retrospective evaluation of polymyxin activity against 6001 Pseudomonas aeruginosa, 150 Achromobacter xylosoxidans and 506 Stenotrophomonas maltophilia CF isolates was initiated. In
Barbara Valentinis et al.
The Journal of biological chemistry, 280(14), 14264-14271 (2005-01-27)
Polymyxin B is a lipopolysaccharide binding antibiotic used to inactivate potential lipopolysaccharide contaminations when evaluating the activity of different agents on innate immune cells. We report that polymyxin B is able to induce directly in monocyte-derived human dendritic cells (DCs)
A Bonura et al.
Allergy, 68(3), 297-303 (2013-01-22)
The major allergens in Parietaria pollen, Par j 1 and Par j 2, have been identified as lipid transfer proteins. The family of the Par j 1 allergens is composed of two isoforms, which differ by the presence of a
Kensuke Nakamura et al.
The American journal of emergency medicine, 31(5), 893-893 (2013-02-13)
Septic patients often have low cardiac output. Some of them present severe cardiac dysfunction such as septic cardiomyopathy. However, no well-known and effective treatment for septic cardiomyopathy exists. The effect of endotoxin adsorption by polymyxin B–immobilized fiber column–direct hemoperfusion (PMX-DHP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service