Skip to Content
Merck
All Photos(1)

Key Documents

P2495

Sigma-Aldrich

Phalloidin, Coumarin Labeled

for actin detection

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H75N11O15S2
Molecular Weight:
1278.45
UNSPSC Code:
12352204
PubChem Substance ID:

form

solid

solubility

methanol: 1 mg/mL, clear, colorless to light yellow-green

storage temp.

−20°C

InChI

1S/C62H75N11O15S2/c1-8-71(9-2)36-18-19-38-30(3)50(61(86)88-46(38)22-36)34-14-16-35(17-15-34)73-49(77)25-47(60(73)85)89-21-20-48(76)63-29-62(7,87)26-43-55(81)64-32(5)53(79)70-51(33(6)74)57(83)68-44-28-90-58-40(39-12-10-11-13-41(39)69-58)24-42(54(80)67-43)66-52(78)31(4)65-56(82)45-23-37(75)27-72(45)59(44)84/h10-19,22,31-33,37,42-45,47,51,69,74-75,87H,8-9,20-21,23-29H2,1-7H3,(H,63,76)(H,64,81)(H,65,82)(H,66,78)(H,67,80)(H,68,83)(H,70,79)/t31-,32-,33-,37-,42-,43-,44-,45-,47?,51+,62+/m0/s1

InChI key

GNSRKMXDGGXWDM-AZPABDIKSA-N

Application

A fluorescent phallotoxin which may be used to identify filamentous actin.

Preparation Note

From phalloidin isolated from Amanita phalloides mushrooms

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Padmanabhan et al.
Endocrinology, 151(2), 595-605 (2009-12-08)
Polycystic ovary syndrome (PCOS) is the most common endocrinopathy of reproductive-aged women and is exacerbated by obesity. Exposure of ewes to excess testosterone (T) from d 30-90 of gestation culminates in anovulation, functional hyperandrogenism, LH excess, and polyfollicular ovaries, features
Steve Dos-Santos Carvalho et al.
eLife, 9 (2020-01-08)
Dynamic mechanical interactions between adhesion complexes and the cytoskeleton are essential for axon outgrowth and guidance. Whether planar cell polarity (PCP) proteins, which regulate cytoskeleton dynamics and appear necessary for some axon guidance, also mediate interactions with membrane adhesion is
Shanhui Ren et al.
PLoS pathogens, 16(6), e1008514-e1008514 (2020-06-02)
Deoxyribonucleic acid (DNA) damage response (DDR) is the fundamental cellular response for maintaining genomic integrity and suppressing tumorigenesis. The activation of ataxia telangiectasia-mutated (ATM) kinase is central to DNA double-strand break (DSB) for maintaining host-genome integrity in mammalian cells. Oncolytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service