Skip to Content
Merck
All Photos(1)

Key Documents

P1326

Sigma-Aldrich

Prostaglandin E1-[5,6-3H(N)]

aqueous ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H35O5
CAS Number:
Molecular Weight:
355.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

aqueous ethanol solution

extent of labeling

25-60 Ci/mmol

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@@H](O)\C=C\[C@@H]1[C@@H](O)CC(=O)[C@@H]1CCCCCCC(O)=O

Packaging

Packaged under Nitrogen

Physical form

Ethanol: water (70:30) solution

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 2 - STOT SE 3

Target Organs

Blood, Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service