Skip to Content
Merck
All Photos(1)

Key Documents

P105

Sigma-Aldrich

(±)-PPHT hydrochloride

≥98% (HPLC), solid

Synonym(s):

(±)-2-(N-Phenethyl-N-propyl)amino-5-hydroxytetralin hydrochloride, N-0434

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO · HCl
CAS Number:
Molecular Weight:
345.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: ≥10 mg/mL
H2O: ≥2 mg/mL

SMILES string

Cl[H].CCCN(CCc1ccccc1)C2CCc3c(O)cccc3C2

InChI

1S/C21H27NO.ClH/c1-2-14-22(15-13-17-7-4-3-5-8-17)19-11-12-20-18(16-19)9-6-10-21(20)23;/h3-10,19,23H,2,11-16H2,1H3;1H

InChI key

XWLCIDLCEZAOEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A series of new dopamine (DA) receptor agonists, of the 2-aminotetralin group, i.e. N-0434, N-0437 and N-0734 were investigated in both in vivo and in vitro pharmacological test systems. In vivo, the reversal of the gamma-butyrolactone-induced increase in rat central DOPA biosynthesis rate was taken as a measure of presynaptic activity. The homovanillic acid (HVA) decrease, after intraperitoneal and after oral administration of the drugs was also taken as a measure of presynaptic activity. Postsynaptic activity was measured in two behavioural models, i.e. reserpine reversal and stereotypy induction. The effects of (±)-PPHT (N-0434) these drugs on noradrenaline and dopamine turnover (alpha-MpT method) were studied in addition. The results indicate that all three compounds N-0434 ((±)-PPHT), N-0437 and N-0734 are potent and selective DA agonists that lack significant alpha 2 activity.
Potent D2 dopamine receptor agonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Díaz-Véliz et al.
Pharmacology, biochemistry, and behavior, 62(1), 21-29 (1999-02-11)
The influence of the hormonal condition on the reactivity of central dopamine (DA) receptors was studied in male and in intact and ovariectomized (OVX) female rats. They were injected with selective DA agonists, acting either on D1 (SKF 38393, 2.5
B Kanyicska et al.
Endocrinology, 138(8), 3141-3153 (1997-08-01)
Endothelin-1 (ET-1) inhibits PRL secretion from cultured rat lactotrophs. However, ET-1 stimulates PRL secretion after cultured lactotrophs have been exposed for 48 h to dopamine or D2 dopamine agonists. In the present study, we have used cell-attached and inside-out patch
M E Meyer et al.
Pharmacology, biochemistry, and behavior, 44(4), 865-868 (1993-04-01)
The effects of a dopamine agonist, (+/-)-2-(N-penylethyl-N-propyl)amino-5- hydroxytetralin (N-0434) (SC doses of 0.00, 0.01, 0.1, and 1.0 mg/kg) were tested in rats for 120 min in an activity monitor. The durations in seconds of horizontal locomotor time, rearing time, stereotypy
F J Bosker et al.
European journal of pharmacology, 163(2-3), 319-326 (1989-04-25)
Partial purification of the dopamine D-2 receptor from bovine striatum, solubilized in the presence of 1% digitonin, was obtained by chromatography on wheat germ lectin agarose. The preparation was purified approximately 10-fold. The stability of the receptor preparation was considerably
A Mao et al.
Life sciences, 59(21), PL317-PL324 (1996-01-01)
The concentrations of endogenous ligands generally remain in a bounded range around a basal level, a manifestation of control. The dopaminergic system is an excellent example of a control system in which a negative feedback signal is associated with receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service