Skip to Content
Merck
All Photos(1)

Key Documents

P0690

Sigma-Aldrich

Pectinase from Aspergillus niger

BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)

Synonym(s):

Poly-(1,4-α-D-galacturonide) glycanohydrolase, Polygalacturonase solution from Aspergillus niger

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352200

product line

BioReagent

form

aqueous glycerol solution

specific activity

≥5 units/mg protein (Lowry)

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Pectinase P0690 is a natural pectinase (polygalacturonase) produced by Aspergillus niger. This enzyme breaks down pectin found in the middle lamella of plant cell walls. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation.
Used in plant protoplast preparation to digest cell wall prior to organelle isolation.

Unit Definition

One unit will liberate 1.0 μmole of galacturonic acid from polygalacturonic acid per min at pH 4.0 at 25 °C.

Physical form

Solution in 40% glycerol

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincenzo Lionetti et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(2), 616-621 (2010-01-19)
Plant cell walls represent an abundant, renewable source of biofuel and other useful products. The major bottleneck for the industrial scale-up of their conversion to simple sugars (saccharification), to be subsequently converted by microorganisms into ethanol or other products, is
Sina Pricelius et al.
Journal of agricultural and food chemistry, 57(3), 1006-1012 (2009-02-05)
Attractive color is one of the most important sensory characteristics of fruit and berry products, and elderberry juice is widely used as natural colorant. When pectinase preparations were used in the production of elderberry juice for clarification, a concomitant decrease
Sara Posé et al.
Journal of experimental botany, 64(12), 3803-3815 (2013-07-23)
Antisense-mediated down-regulation of the fruit-specific polygalacturonase (PG) gene FaPG1 in strawberries (Fragaria×ananassa Duch.) has been previously demonstrated to reduce fruit softening and to extend post-harvest shelf life, despite the low PG activity detected in this fruit. The improved fruit traits
Mi Wei et al.
Bioresource technology, 146, 549-555 (2013-08-27)
A novel method utilizing microbial treatment for cleaner production of diosgenin from Dioscorea zingiberensis C.H. Wright (DZW) was presented. A new Bacillus pumilus HR19, which has the great ability to secrete pectinase, was screened and applied in the microbial treatment.
Isabelle Benoit et al.
BMC genomics, 13, 321-321 (2012-07-21)
Pectins are diverse and very complex biomolecules and their structure depends on the plant species and tissue. It was previously shown that derivatives of pectic polymers and oligosaccharides from pectins have positive effects on human health. To obtain specific pectic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service