Skip to Content
Merck
All Photos(1)

Key Documents

P0622

Sigma-Aldrich

PET-cGMP

≥98% (HPLC), solid

Synonym(s):

β-Phenyl-1,N2-ethenoguanosine 3′,5′-monophosphate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N5O7PNa
CAS Number:
Molecular Weight:
467.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

packaging

pkg of 10 μmol

color

white

solubility

H2O: ~10 mmol

shipped in

dry ice

storage temp.

−70°C

SMILES string

[Na+].O[C@@H]1[C@@H]2OP([O-])(=O)OC[C@H]2O[C@H]1n3cnc4C(=O)N5C=C(NC5=Nc34)c6ccccc6

InChI

1S/C18H16N5O7P.Na/c24-13-14-11(7-28-31(26,27)30-14)29-17(13)23-8-19-12-15(23)21-18-20-10(6-22(18)16(12)25)9-4-2-1-3-5-9;/h1-6,8,11,13-14,17,24H,7H2,(H,20,21)(H,26,27);/q;+1/p-1/t11-,13-,14-,17-;/m1./s1

InChI key

IGIOCJJIGJLGKR-TZNCIMHNSA-M

Biochem/physiol Actions

cGMP-dependent protein kinase (PKG I and PKG II) activator. Potent membrane permeant activator of both isozymes I α and I β of cGMP-dependent protein kinase. Common stimulators such as 8-Br-cGMP or 8-pCPT-cGMP mainly activate kinase subtype Iα.

Features and Benefits

This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

James C Campbell et al.
ACS chemical biology, 12(9), 2388-2398 (2017-08-10)
Cyclic GMP analogs, 8-Br, 8-pCPT, and PET-cGMP, have been widely used for characterizing cellular functions of cGMP-dependent protein kinase (PKG) I and II isotypes. However, interpreting results obtained using these analogs has been difficult due to their low isotype specificity.
Stephanie D Nofal et al.
mBio, 12(1) (2021-01-28)
Guanylyl cyclases (GCs) synthesize cyclic GMP (cGMP) and, together with cyclic nucleotide phosphodiesterases, are responsible for regulating levels of this intracellular messenger which mediates myriad functions across eukaryotes. In malaria parasites (Plasmodium spp), as well as their apicomplexan and ciliate
Matthew Tcheng et al.
Blood, 137(25), 3518-3532 (2021-03-16)
Acute myeloid leukemia (AML) cells have an atypical metabolic phenotype characterized by increased mitochondrial mass, as well as a greater reliance on oxidative phosphorylation and fatty acid oxidation (FAO) for survival. To exploit this altered metabolism, we assessed publicly available

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service