These proteins bind to the catalytic subunit of cyclic-AMP dependent protein kinase, displacing the regulatory subunit and inhibiting phosphorylating activity. The cyclic-AMP binding capacity of the regulatory subunit is enhanced. Many other proteins exhibit this enhancement ability, too (Brostrom, C.O. and Kon, C., Anal. Biochem., 58, 459 [1974]).
Packaging
Package size based on protein content.
Unit Definition
One unit will transfer 1.0 picomole (10-12 mol) of phosphate from γ-32P-ATP to hydrolyzed and partially dephosphorylated casein (Product No. C 4765) per min at pH 6.5 at 30 °C.
Physical form
Supplied as lyophilized powder containing 30% glycerophosphate and 20% EDTA.
Preparation Note
Fractionated essentially by procedure of Walsh, D.A., et al., J. Biol. Chem., 246, 1977 (1971).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.