Skip to Content
Merck
All Photos(1)

Key Documents

P0248

Sigma-Aldrich

PNU-37883A

≥98% (HPLC), powder

Synonym(s):

N-(1-Adamantyl)-N′-cyclohexyl-4-morpholinecarboxamidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H36ClN3O
CAS Number:
Molecular Weight:
381.98
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: ~23 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Cl.C1CCC(CC1)\N=C(/NC23CC4CC(CC(C4)C2)C3)N5CCOCC5

Gene Information

human ... KCNJ8(3764)

Biochem/physiol Actions

Selective inhibitor of Kir6.1/SUR2B channels.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kovalev et al.
British journal of pharmacology, 141(5), 867-873 (2004-02-06)
1. In this study, we have used Kir6.1/Kir6.2 chimeric proteins and current recordings to investigate the molecular basis of PNU-37883A inhibition of cloned K(ATP) channels. 2. Rat Kir6.1, Kir6.2 and Kir6.1/Kir6.2 chimeras were co-expressed with either SUR2B or SUR1, following
Bobby D Nossaman et al.
Journal of cardiovascular pharmacology, 50(4), 358-366 (2007-12-01)
The free radical peroxynitrite (ONOO-) is formed in biological systems from the reaction of nitric oxide (NO) with superoxide (O2-) and can react with protein and nonprotein thiol groups to produce tissue injury. However, these pathologic actions of (ONOO-) may
Mariko Yamanaka et al.
International immunopharmacology, 2(13-14), 1957-1964 (2002-12-20)
It is suggested that attenuation of the renal kallikrein-kinin system (KKS) involved the development of hypertension in young spontaneously hypertensive rats (SHR). In the present study, a comparison was made between young SHR and Wistar Kyoto rats (WKY) to examine
Salma Toma Hanna et al.
European journal of pharmacology, 515(1-3), 34-42 (2005-05-17)
Kir6.1 subunit is one of the pore-forming components of K(ATP) channel complex. The endogenous modulation of Kir6.1 subunit function has been largely unknown. Whether acetylcholine modulated the function of Kir6.1 subunit stably expressed in human embryonic kidney (HEK-293) cells was
Noriyoshi Teramoto et al.
European journal of pharmacology, 506(1), 1-7 (2004-12-14)
Kinetic studies of U-37883A (4-morpholinecarboximidine-N-1-adamantyl-N'-cyclohexyl-hydrochloride), a vascular ATP-sensitive K+ channel (KATP channel) blocker, were performed on pig urethral myocytes to investigate inhibitory effects on large-conductance intracellular Ca2+ -sensitive K+ channels (i.e., BKCa channels; 225 pS K+ channels) by use of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service