Skip to Content
Merck
All Photos(1)

Key Documents

P0233

Sigma-Aldrich

Anti-Potassium Channel Kv4.2 (Shal1; RK5; Kcnd2) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 ML
$1,150.00

$1,150.00


Check Cart for Availability


Select a Size

Change View
0.2 ML
$1,150.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$1,150.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, human

technique(s)

western blot (chemiluminescent): 1:200-1:300

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCND2(3751)
rat ... Kcnd2(65180)

Specificity

Recognizes full-length Kv4.2 protein.

Immunogen

synthetic peptide corresponding to amino acids 454-469 of rat potassium channel Kv4.2 (with additional N-terminal cysteine). The epitope is identical in human and highly homologous in mouse.

Application

western blot (chemiluminescent): 1:200-1:300

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.025% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine Lotteau et al.
Journal of the American Heart Association, 10(17), e019273-e019273 (2021-09-03)
Background Sodium-calcium (Ca2+) exchanger isoform 1 (NCX1) is the dominant Ca2+ efflux mechanism in cardiomyocytes and is critical to maintaining Ca2+ homeostasis during excitation-contraction coupling. NCX1 activity has been implicated in the pathogenesis of cardiovascular diseases, but a lack of
Jae Hyung Cho et al.
JCI insight, 3(19) (2018-10-05)
Sudden death is the most common mode of exodus in patients with heart failure and preserved ejection fraction (HFpEF). Cardiosphere-derived cells (CDCs) reduce inflammation and fibrosis in a rat model of HFpEF, improving diastolic function and prolonging survival. We tested
Jia-Hua Hu et al.
International journal of molecular sciences, 21(16) (2020-08-23)
The subthreshold, transient A-type K+ current is a vital regulator of the excitability of neurons throughout the brain. In mammalian hippocampal pyramidal neurons, this current is carried primarily by ion channels comprising Kv4.2 α-subunits. These channels occupy the somatodendritic domains
Jia-Hua Hu et al.
Nature communications, 11(1), 1567-1567 (2020-03-29)
Voltage-gated K+ channels function in macromolecular complexes with accessory subunits to regulate brain function. Here, we describe a peptidyl-prolyl cis-trans isomerase NIMA-interacting 1 (Pin1)-dependent mechanism that regulates the association of the A-type K+ channel subunit Kv4.2 with its auxiliary subunit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service