Skip to Content
Merck
All Photos(1)

Key Documents

P0046

Sigma-Aldrich

Pyocyanin

from Pseudomonas aeruginosa, ≥98% (HPLC), solid, redox-active phenazine

Synonym(s):

5-Methyl-1(5H)-phenazinone, Pyocyanine, Sanasin, Sanazin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$221.00
25 MG
$970.00

$221.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$221.00
25 MG
$970.00

About This Item

Empirical Formula (Hill Notation):
C13H10N2O
CAS Number:
Molecular Weight:
210.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$221.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Pyocyanin, from Pseudomonas aeruginosa, ≥98% (HPLC)

biological source

Pseudomonas aeruginosa

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble
acetone: soluble
chloroform: soluble
ethanol: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

CN1c2ccccc2N=C3C(=O)C=CC=C13

InChI

1S/C13H10N2O/c1-15-10-6-3-2-5-9(10)14-13-11(15)7-4-8-12(13)16/h2-8H,1H3

InChI key

YNCMLFHHXWETLD-UHFFFAOYSA-N

Application

Pyocyanin has been used as a biomarker to detect Pseudomonas aeruginosa.[1][2][3]

Biochem/physiol Actions

Pyocyanin, a blue-green pigment belonging to phenazine pigments, is a redox-active phenazine. Pyocyanin is an electron receptor, which stimulates redox cycling in bacteria, liver cells, and human epithlial cell lines.
Pyocyanin, a blue-green pigment belonging to phenazine pigments, is a redox-active phenazine. Pyocyanin is an electron receptor, which stimulates redox cycling in bacteria, liver cells, and human epithlial cell lines. Pyocyanin enhances the oxidative metabolism, which in turn increases the formation of intracellular reactive oxygen species (ROS) via reduction of NADPH. Pyocyanin also increases the release of IL-8 by airway epithelial cells both in vitro and in vivo. This involves signal transduction pathways that include oxidants, protein tyrosine kinases, and MAP kinases. IL-8 secretion by these cells is in synergy with inflammatory cytokines. Pyocyanin has been shown to accelerate neutrophil apoptosis in vitro, resulting in resolution of acute inflammation, which is beneficial for bacteria survival.

Preparation Note

For the preparation of an aqueous solution, dissolve pyocyanin in ethanol using 10% of the final required volume and then add distilled water to complete the volume.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast selective detection of pyocyanin using cyclic voltammetry
Alatraktchi F, et al.
Sensors, 16(3), 408-408 (2016)
Mout De Vrieze et al.
Frontiers in microbiology, 11, 857-857 (2020-05-20)
For plants, the advantages of associating with beneficial bacteria include plant growth promotion, reduction of abiotic and biotic stresses and enhanced protection against various pests and diseases. Beneficial bacteria rightly equipped for successful plant colonization and showing antagonistic activity toward
Yangmei Li et al.
Microbial biotechnology, 13(4), 1132-1144 (2020-04-05)
Pf prophages are ssDNA filamentous prophages that are prevalent among various Pseudomonas aeruginosa strains. The genomes of Pf prophages contain not only core genes encoding functions involved in phage replication, structure and assembly but also accessory genes. By studying the
Mariola Andrejko et al.
Acta biochimica Polonica, 66(1), 91-100 (2019-03-05)
Thermally induced unfolding and renaturation capability of alkaline proteases (AprA) of three Pseudomonas aeruginosa strains, i.e. ATCC 27853 and two clinical isolates, was examined. Sequence analyses demonstrated a high level of aprA genes identity (99.24-99.8%) in these bacterial strains. The
Theerthankar Das et al.
Frontiers in microbiology, 8, 2429-2429 (2018-01-10)
Pyocyanin secreted by

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

  1. ①試薬の保存温度として―80℃は不適切でしょうか。 ②純水に溶解後の保存方法として、冷蔵保存することは可能でしょうか。 宜しくお願いいたします。

    1 answer
    1. The recommended storage temperature for this product is for –20°C. The stability of this product at –80°C has not been evaluated. This compound is not directly soluble in water. However, product R9532, Pyocyanin, Ready Made Solution, is a 5 mg/mL solution in DMSO. This item has a general shelf live of 2 years at –20°C. The prepared solution is not stable when stored at refrigerated temperatures. Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/429/027/p0046dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service