Skip to Content
Merck
All Photos(1)

Key Documents

P0031

Sigma-Aldrich

Protopanaxadiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H52O3
CAS Number:
Molecular Weight:
460.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

form

powder

color

white

solubility

methanol: 5 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

C[C@@]12CC[C@@]3([H])C(C)(C)[C@@H](O)CC[C@]3(C)[C@@]1([H])C[C@@H](O)[C@@]4([H])[C@]2(CC[C@]4([H])[C@@](CCC=C(C)C)(C)O)C

InChI

1S/C30H52O3/c1-19(2)10-9-14-30(8,33)20-11-16-29(7)25(20)21(31)18-23-27(5)15-13-24(32)26(3,4)22(27)12-17-28(23,29)6/h10,20-25,31-33H,9,11-18H2,1-8H3/t20-,21+,22-,23+,24-,25-,27-,28+,29+,30-/m0/s1

InChI key

PYXFVCFISTUSOO-HKUCOEKDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A ginsenoside metabolite that has been linked to endothelial cell function via the glucocortoid receptor (GR) and the oestrogen receptor (ER).

Application

Protopanaxadiol [20(R)-protopanaxadiol], a dammarane-type tetracyclic terpene sapogenin, may be used to study its binding to and modulation of endothelial cell function via glucocortoid (GR) and the oestrogen (ER) receptors. Protopanaxadiol may be used to study its catabolism by cytochrome-P450s. 20(R)-protopanaxadiol may be used in comparative studies versus 20(S)-protopanaxadiol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Zhen et al.
Science China. Life sciences, 58(11), 1099-1110 (2015-11-14)
Panax ginseng C. A. Meyer is an important traditional herb in eastern Asia. It contains ginsenosides, which are primary bioactive compounds with medicinal properties. Although ginseng has been cultivated since at least the Ming dynasty to increase production, cultivated ginseng
Eun-Ah Bae et al.
Biological & pharmaceutical bulletin, 25(1), 58-63 (2002-02-05)
When ginsenoside Rg3 was anaerobically incubated with human fecal microflora, all specimens metabolized ginsenoside Rg3 to ginsenoside Rh2 and protopanaxadiol. The main metabolite was ginsenoside Rh2. 20(S)-ginsenoside Rg3 was quickly transformed to 20(S)-ginsenoside Rh2 or 20(S)-protopanaxadiol in an amount 19-fold
Xin Jin et al.
Fitoterapia, 84, 64-71 (2012-09-26)
As with many other anti-cancer agents, 20(S)-protopanaxadiol (PPD) has a low oral absorption. In this study, in order to improve the oral bioavailability of PPD, the cubic nanoparticles that it contains were used to enhance absorption. Therefore, the cubic nanoparticle
Liang Li et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(3), 472-483 (2010-12-09)
20(S)-Protopanaxadiol (PPD, 1) is one of the aglycones of the ginsenosides and has a wide range of pharmacological activities. At present, PPD has progressed to early clinical trials as an antidepressant. In this study, its fate in mixed human liver
Alain Gulenga Musende et al.
Anti-cancer drugs, 23(5), 543-552 (2012-04-07)
This study focuses on determining the pharmacokinetics, biodistribution, and efficacy of the ginsenoside aglycone protopanaxadiol (aPPD) administered as a single agent in a novel oral dosage formulation. To obtain these data and to characterize the stability of aPPD, appropriate analytical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service